Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178034909-178035458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM153C
Entrez Gene ID
653316
Gene Name
family with sequence similarity 153, member C
Gene Aliases
NY-REN-7-like
UniGene
Hs.652193
Gene Chromosome Location
Chr.5: 178006966-178049087 on Build GRCh38
Transcripts
NR_038353.1

SNP Detail

SNP ID Chromosome Location
rs563029985 Chr.5: 178034942-178034942 on Build GRCh38
rs887509387 Chr.5: 178034940-178034940 on Build GRCh38
rs548805916 Chr.5: 178035014-178035014 on Build GRCh38
rs771728418 Chr.5: 178035120-178035120 on Build GRCh38
rs775239870 Chr.5: 178035136-178035136 on Build GRCh38
rs534078519 Chr.5: 178035076-178035076 on Build GRCh38
rs770769004 Chr.5: 178035101-178035101 on Build GRCh38
rs774434233 Chr.5: 178035102-178035102 on Build GRCh38
rs777408568 Chr.5: 178035053-178035053 on Build GRCh38
rs763384042 Chr.5: 178035160-178035160 on Build GRCh38
rs570772645 Chr.5: 178035201-178035201 on Build GRCh38
rs552458436 Chr.5: 178035138-178035138 on Build GRCh38
rs537468987 Chr.5: 178035289-178035289 on Build GRCh38
rs139377481 Chr.5: 178034970-178034970 on Build GRCh38
rs760209845 Chr.5: 178035137-178035137 on Build GRCh38
rs745316756 Chr.5: 178035115-178035115 on Build GRCh38
rs1014697080 Chr.5: 178034991-178034991 on Build GRCh38
rs1004613414 Chr.5: 178034972-178034972 on Build GRCh38
rs555863711 Chr.5: 178035366-178035366 on Build GRCh38
rs566718770 Chr.5: 178035039-178035039 on Build GRCh38
rs773449509 Chr.5: 178035152-178035152 on Build GRCh38
rs896288018 Chr.5: 178035041-178035041 on Build GRCh38
rs763852927 Chr.5: 178035146-178035146 on Build GRCh38
rs530123625 Chr.5: 178034986-178034986 on Build GRCh38

View Full Product Details