Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178041700-178042249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM153C
Entrez Gene ID
653316
Gene Name
family with sequence similarity 153, member C
Gene Aliases
NY-REN-7-like
UniGene
Hs.652193
Gene Chromosome Location
Chr.5: 178006966-178049087 on Build GRCh38
Transcripts
NR_038353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1311029 c.382G>A p.E128K Chr.5: 178041710-178041710 on Build GRCh38
3744797 c.202A>G p.M68V Chr.5: 178041761-178041761 on Build GRCh38
1137316 c.458C>T p.P153L Chr.5: 178042217-178042217 on Build GRCh38
3614595 c.142G>A p.E48K Chr.5: 178041701-178041701 on Build GRCh38
345738 c.209A>T p.Q70L Chr.5: 178042199-178042199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201786531 Chr.5: 178041758-178041758 on Build GRCh38
rs13165552 Chr.5: 178041868-178041868 on Build GRCh38
rs772508355 Chr.5: 178042176-178042176 on Build GRCh38
rs765609069 Chr.5: 178041768-178041768 on Build GRCh38
rs535110605 Chr.5: 178041837-178041837 on Build GRCh38
rs13165684 Chr.5: 178041917-178041917 on Build GRCh38
rs958694753 Chr.5: 178042049-178042049 on Build GRCh38
rs4613734 Chr.5: 178041719-178041719 on Build GRCh38
rs1025925278 Chr.5: 178041973-178041973 on Build GRCh38
rs776754450 Chr.5: 178042205-178042205 on Build GRCh38
rs764263059 Chr.5: 178041720-178041720 on Build GRCh38
rs200505161 Chr.5: 178042179-178042179 on Build GRCh38
rs780733800 Chr.5: 178041808-178041808 on Build GRCh38
rs528446701 Chr.5: 178042158-178042158 on Build GRCh38
rs766169446 Chr.5: 178042225-178042225 on Build GRCh38
rs757677888 Chr.5: 178041913-178041913 on Build GRCh38
rs549083636 Chr.5: 178041788-178041788 on Build GRCh38
rs199762283 Chr.5: 178042150-178042150 on Build GRCh38
rs537711525 Chr.5: 178042175-178042175 on Build GRCh38
rs562827882 Chr.5: 178042104-178042104 on Build GRCh38
rs1011565480 Chr.5: 178042080-178042080 on Build GRCh38
rs373861982 Chr.5: 178042218-178042218 on Build GRCh38
rs923528292 Chr.5: 178041899-178041899 on Build GRCh38
rs549437511 Chr.5: 178042130-178042130 on Build GRCh38
rs370121752 Chr.5: 178042123-178042123 on Build GRCh38
rs576985620 Chr.5: 178042070-178042070 on Build GRCh38
rs544050995 Chr.5: 178042071-178042071 on Build GRCh38
rs761212136 Chr.5: 178042198-178042198 on Build GRCh38
rs13165704 Chr.5: 178041945-178041945 on Build GRCh38
rs978020181 Chr.5: 178041858-178041858 on Build GRCh38
rs530959947 Chr.5: 178041731-178041731 on Build GRCh38
rs369856765 Chr.5: 178041754-178041754 on Build GRCh38
rs377389054 Chr.5: 178041772-178041772 on Build GRCh38
rs907056388 Chr.5: 178041984-178041984 on Build GRCh38
rs758285767 Chr.5: 178041787-178041787 on Build GRCh38
rs13166036 Chr.5: 178042122-178042122 on Build GRCh38
rs11958715 Chr.5: 178041836-178041836 on Build GRCh38
rs751549819 Chr.5: 178041793-178041793 on Build GRCh38
rs13188778 Chr.5: 178041999-178041999 on Build GRCh38
rs942346914 Chr.5: 178041939-178041939 on Build GRCh38
rs13188629 Chr.5: 178041919-178041919 on Build GRCh38
rs114660361 Chr.5: 178041963-178041963 on Build GRCh38
rs13188779 Chr.5: 178042001-178042001 on Build GRCh38
rs754914694 Chr.5: 178041796-178041796 on Build GRCh38
rs13188791 Chr.5: 178042017-178042017 on Build GRCh38
rs769088897 Chr.5: 178042201-178042201 on Build GRCh38
rs190921663 Chr.5: 178042061-178042061 on Build GRCh38
rs13173211 Chr.5: 178042019-178042019 on Build GRCh38
rs867663158 Chr.5: 178042010-178042010 on Build GRCh38
rs765501673 Chr.5: 178042217-178042217 on Build GRCh38
rs747716270 Chr.5: 178042145-178042145 on Build GRCh38
rs142660284 Chr.5: 178041941-178041941 on Build GRCh38
rs761263389 Chr.5: 178041793-178041793 on Build GRCh38
rs553649847 Chr.5: 178041864-178041864 on Build GRCh38
rs571871772 Chr.5: 178041908-178041908 on Build GRCh38
rs564599628 Chr.5: 178042062-178042062 on Build GRCh38
rs762047376 Chr.5: 178042215-178042215 on Build GRCh38

View Full Product Details