Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 108761845-108762424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP2
Entrez Gene ID
5903
Gene Name
RAN binding protein 2
Gene Aliases
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2
UniGene
Hs.199561
Gene Chromosome Location
Chr.2: 108719446-109842301 on Build GRCh38
Transcripts
NM_006267.4 XM_005264002.2 XM_005264003.2 XM_005264004.2 XM_005264005.4 XM_005264007.2 XM_011511575.2 XM_011511576.2 XM_011511578.2 XM_017004623.1 XM_017004624.1 XM_017004625.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1494473 c.2603-2A>T p.? Chr.2: 108762099-108762099 on Build GRCh38
7127672 c.2615G>C p.G872A Chr.2: 108762113-108762113 on Build GRCh38
714825 c.2603T>G p.V868G Chr.2: 108762101-108762101 on Build GRCh38
1744235 c.2623G>C p.V875L Chr.2: 108762121-108762121 on Build GRCh38
6323735 c.2603-1G>T p.? Chr.2: 108762100-108762100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2949969 Chr.2: 108762034-108762034 on Build GRCh38
rs894569873 Chr.2: 108762019-108762019 on Build GRCh38
rs3872500 Chr.2: 108761915-108761915 on Build GRCh38
rs972225570 Chr.2: 108762046-108762047 on Build GRCh38
rs761734747 Chr.2: 108762247-108762247 on Build GRCh38
rs945884996 Chr.2: 108762270-108762270 on Build GRCh38
rs746121307 Chr.2: 108762204-108762204 on Build GRCh38
rs765287306 Chr.2: 108762049-108762049 on Build GRCh38
rs775613285 Chr.2: 108762085-108762085 on Build GRCh38
rs773490660 Chr.2: 108762051-108762051 on Build GRCh38
rs2433787 Chr.2: 108762024-108762024 on Build GRCh38
rs762835776 Chr.2: 108762089-108762089 on Build GRCh38
rs781677563 Chr.2: 108762200-108762200 on Build GRCh38
rs770023583 Chr.2: 108762214-108762214 on Build GRCh38
rs747413002 Chr.2: 108762054-108762054 on Build GRCh38
rs2912832 Chr.2: 108762098-108762098 on Build GRCh38
rs774276043 Chr.2: 108762241-108762241 on Build GRCh38
rs771369374 Chr.2: 108762062-108762063 on Build GRCh38
rs904399751 Chr.2: 108761935-108761935 on Build GRCh38
rs755777927 Chr.2: 108762116-108762116 on Build GRCh38
rs533200484 Chr.2: 108762232-108762232 on Build GRCh38
rs3932816 Chr.2: 108761953-108761953 on Build GRCh38
rs1014372149 Chr.2: 108762028-108762028 on Build GRCh38
rs545206357 Chr.2: 108762261-108762261 on Build GRCh38
rs1034326177 Chr.2: 108761995-108761995 on Build GRCh38
rs879163615 Chr.2: 108762372-108762372 on Build GRCh38
rs775754217 Chr.2: 108762086-108762086 on Build GRCh38
rs757056887 Chr.2: 108762099-108762099 on Build GRCh38
rs2557903 Chr.2: 108762091-108762091 on Build GRCh38
rs1041654908 Chr.2: 108761910-108761910 on Build GRCh38
rs562634196 Chr.2: 108762228-108762228 on Build GRCh38
rs777011613 Chr.2: 108762075-108762075 on Build GRCh38
rs544495075 Chr.2: 108762157-108762157 on Build GRCh38
rs200515712 Chr.2: 108762123-108762123 on Build GRCh38
rs368326522 Chr.2: 108761892-108761892 on Build GRCh38
rs555757226 Chr.2: 108762062-108762062 on Build GRCh38
rs763114639 Chr.2: 108762225-108762225 on Build GRCh38
rs373286631 Chr.2: 108762113-108762113 on Build GRCh38
rs747464401 Chr.2: 108762192-108762192 on Build GRCh38
rs2433711 Chr.2: 108761879-108761879 on Build GRCh38
rs778449584 Chr.2: 108762187-108762187 on Build GRCh38
rs2433710 Chr.2: 108762014-108762014 on Build GRCh38
rs754409615 Chr.2: 108762158-108762158 on Build GRCh38
rs2556263 Chr.2: 108762080-108762080 on Build GRCh38
rs775531921 Chr.2: 108762217-108762217 on Build GRCh38
rs878951886 Chr.2: 108762377-108762377 on Build GRCh38
rs767232631 Chr.2: 108762106-108762106 on Build GRCh38
rs2912833 Chr.2: 108762004-108762004 on Build GRCh38
rs764119940 Chr.2: 108762092-108762092 on Build GRCh38
rs771430206 Chr.2: 108762197-108762197 on Build GRCh38

View Full Product Details