Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40610108-40610278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KNL1
Entrez Gene ID
57082
Gene Name
kinetochore scaffold 1
Gene Aliases
AF15Q14, CASC5, CT29, D40, MCPH4, PPP1R55, Spc7, hKNL-1, hSpc105
UniGene
Hs.181855
Gene Chromosome Location
Chr.15: 40594012-40664342 on Build GRCh38
Transcripts
NM_144508.4 NM_170589.4 XM_017022432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3500916 c.211G>A p.E71K Chr.15: 40610258-40610258 on Build GRCh38
699931 c.222G>C p.M74I Chr.15: 40610269-40610269 on Build GRCh38
6620377 c.210G>A p.T70T Chr.15: 40610257-40610257 on Build GRCh38
6077215 c.231G>C p.V77V Chr.15: 40610278-40610278 on Build GRCh38
4630109 c.230T>C p.V77A Chr.15: 40610277-40610277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765368587 Chr.15: 40610159-40610159 on Build GRCh38
rs750490269 Chr.15: 40610176-40610176 on Build GRCh38
rs749028064 Chr.15: 40610201-40610202 on Build GRCh38
rs200583031 Chr.15: 40610222-40610222 on Build GRCh38
rs758343269 Chr.15: 40610226-40610226 on Build GRCh38
rs765519998 Chr.15: 40610211-40610211 on Build GRCh38
rs561154218 Chr.15: 40610162-40610162 on Build GRCh38
rs956708293 Chr.15: 40610164-40610164 on Build GRCh38
rs766473801 Chr.15: 40610220-40610220 on Build GRCh38
rs955136949 Chr.15: 40610166-40610166 on Build GRCh38
rs1032770705 Chr.15: 40610153-40610153 on Build GRCh38
rs770849722 Chr.15: 40610250-40610253 on Build GRCh38
rs750540311 Chr.15: 40610214-40610214 on Build GRCh38
rs762151589 Chr.15: 40610203-40610203 on Build GRCh38
rs763179214 Chr.15: 40610215-40610215 on Build GRCh38

View Full Product Details