Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55160140-55160294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFR
Entrez Gene ID
1956
Gene Name
epidermal growth factor receptor
Gene Aliases
ERBB, ERBB1, HER1, NISBD2, PIG61, mENA
UniGene
Hs.488293
Gene Chromosome Location
Chr.7: 55019032-55207338 on Build GRCh38
Transcripts
NM_005228.3 NM_201282.1 NM_201284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6471959 c.1392_1397delAGGAAA p.G465_N466delGN Chr.7: 55160232-55160237 on Build GRCh38
4604310 c.1376G>C p.G459A Chr.7: 55160216-55160216 on Build GRCh38
1090873 c.1310C>A p.S437Y Chr.7: 55160150-55160150 on Build GRCh38
6957936 c.1335A>G p.I445M Chr.7: 55160175-55160175 on Build GRCh38
5757060 c.1431G>A p.W477* Chr.7: 55160271-55160271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009449079 Chr.7: 55160241-55160241 on Build GRCh38
rs1021252393 Chr.7: 55160265-55160265 on Build GRCh38
rs755147377 Chr.7: 55160211-55160211 on Build GRCh38
rs753269876 Chr.7: 55160218-55160218 on Build GRCh38
rs866211550 Chr.7: 55160247-55160247 on Build GRCh38
rs199754312 Chr.7: 55160235-55160235 on Build GRCh38
rs765091640 Chr.7: 55160165-55160165 on Build GRCh38
rs372990493 Chr.7: 55160173-55160173 on Build GRCh38
rs751667594 Chr.7: 55160192-55160192 on Build GRCh38
rs779726192 Chr.7: 55160262-55160262 on Build GRCh38
rs1025827299 Chr.7: 55160193-55160193 on Build GRCh38
rs1032006770 Chr.7: 55160171-55160171 on Build GRCh38
rs771736232 Chr.7: 55160253-55160253 on Build GRCh38
rs746521110 Chr.7: 55160267-55160267 on Build GRCh38
rs746763556 Chr.7: 55160230-55160230 on Build GRCh38
rs377567759 Chr.7: 55160191-55160191 on Build GRCh38
rs146711874 Chr.7: 55160214-55160214 on Build GRCh38
rs200592648 Chr.7: 55160222-55160222 on Build GRCh38
rs750541010 Chr.7: 55160182-55160182 on Build GRCh38

View Full Product Details