Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 140734742-140734890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRAF
Entrez Gene ID
673
Gene Name
B-Raf proto-oncogene, serine/threonine kinase
Gene Aliases
B-RAF1, B-raf, BRAF1, NS7, RAFB1
UniGene
Hs.550061
Gene Chromosome Location
Chr.7: 140719331-140924764 on Build GRCh38
Transcripts
NM_004333.4 XM_005250045.2 XM_017012558.1 XM_017012559.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7347329 c.2128-6_2128-5delTT p.? Chr.7: 140734775-140734776 on Build GRCh38
2861420 c.2134G>A p.A712T Chr.7: 140734764-140734764 on Build GRCh38
7347364 c.2128-16_2128-7delCTTTTTTTTT p.? Chr.7: 140734777-140734786 on Build GRCh38
1184859 c.2153C>T p.A718V Chr.7: 140734745-140734745 on Build GRCh38
7346468 c.2128-3T>G p.? Chr.7: 140734773-140734773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777879611 Chr.7: 140734784-140734828 on Build GRCh38
rs202235202 Chr.7: 140734796-140734796 on Build GRCh38
rs374747052 Chr.7: 140734797-140734797 on Build GRCh38
rs151306633 Chr.7: 140734775-140734775 on Build GRCh38
rs769351098 Chr.7: 140734787-140734787 on Build GRCh38
rs60814637 Chr.7: 140734797-140734798 on Build GRCh38
rs397516899 Chr.7: 140734775-140734775 on Build GRCh38
rs753979169 Chr.7: 140734811-140734811 on Build GRCh38
rs760383952 Chr.7: 140734822-140734824 on Build GRCh38
rs748461475 Chr.7: 140734780-140734786 on Build GRCh38
rs199611819 Chr.7: 140734835-140734836 on Build GRCh38
rs368721021 Chr.7: 140734786-140734786 on Build GRCh38
rs772840146 Chr.7: 140734821-140734824 on Build GRCh38
rs750642345 Chr.7: 140734804-140734804 on Build GRCh38
rs886062016 Chr.7: 140734786-140734797 on Build GRCh38
rs767730585 Chr.7: 140734798-140734798 on Build GRCh38
rs767162242 Chr.7: 140734791-140734807 on Build GRCh38
rs745339364 Chr.7: 140734773-140734773 on Build GRCh38
rs766844227 Chr.7: 140734777-140734786 on Build GRCh38
rs373442098 Chr.7: 140734774-140734775 on Build GRCh38
rs753056231 Chr.7: 140734799-140734814 on Build GRCh38
rs397813649 Chr.7: 140734806-140734807 on Build GRCh38
rs575763671 Chr.7: 140734803-140734804 on Build GRCh38
rs529717783 Chr.7: 140734814-140734816 on Build GRCh38
rs771172844 Chr.7: 140734787-140734787 on Build GRCh38
rs758588300 Chr.7: 140734779-140734780 on Build GRCh38
rs376886058 Chr.7: 140734786-140734786 on Build GRCh38
rs727502904 Chr.7: 140734763-140734763 on Build GRCh38
rs772783311 Chr.7: 140734796-140734797 on Build GRCh38
rs55806659 Chr.7: 140734820-140734820 on Build GRCh38
rs745959394 Chr.7: 140734814-140734815 on Build GRCh38
rs755656509 Chr.7: 140734776-140734797 on Build GRCh38
rs397516898 Chr.7: 140734780-140734780 on Build GRCh38
rs779398466 Chr.7: 140734778-140734797 on Build GRCh38
rs61291709 Chr.7: 140734804-140734806 on Build GRCh38
rs754390169 Chr.7: 140734777-140734797 on Build GRCh38
rs370701493 Chr.7: 140734813-140734814 on Build GRCh38
rs757410371 Chr.7: 140734806-140734845 on Build GRCh38
rs758719672 Chr.7: 140734802-140734807 on Build GRCh38
rs377165711 Chr.7: 140734762-140734762 on Build GRCh38
rs778023958 Chr.7: 140734803-140734807 on Build GRCh38
rs764063029 Chr.7: 140734822-140734822 on Build GRCh38
rs766384419 Chr.7: 140734809-140734809 on Build GRCh38
rs201542846 Chr.7: 140734814-140734814 on Build GRCh38
rs751877648 Chr.7: 140734803-140734832 on Build GRCh38
rs776003828 Chr.7: 140734784-140734784 on Build GRCh38
rs758330124 Chr.7: 140734819-140734819 on Build GRCh38
rs886062015 Chr.7: 140734785-140734786 on Build GRCh38
rs760743162 Chr.7: 140734807-140734807 on Build GRCh38
rs145368076 Chr.7: 140734820-140734821 on Build GRCh38
rs535209939 Chr.7: 140734818-140734818 on Build GRCh38
rs749903414 Chr.7: 140734776-140734786 on Build GRCh38
rs761539834 Chr.7: 140734789-140734797 on Build GRCh38
rs886062014 Chr.7: 140734774-140734774 on Build GRCh38
rs774138098 Chr.7: 140734788-140734797 on Build GRCh38
rs397516900 Chr.7: 140734776-140734776 on Build GRCh38
rs760158977 Chr.7: 140734798-140734800 on Build GRCh38
rs956143558 Chr.7: 140734774-140734774 on Build GRCh38
rs370332827 Chr.7: 140734786-140734787 on Build GRCh38
rs535630330 Chr.7: 140734815-140734820 on Build GRCh38
rs200465027 Chr.7: 140734797-140734797 on Build GRCh38
rs556212081 Chr.7: 140734778-140734786 on Build GRCh38
rs768589563 Chr.7: 140734815-140734818 on Build GRCh38
rs368398788 Chr.7: 140734833-140734834 on Build GRCh38
rs376053513 Chr.7: 140734812-140734814 on Build GRCh38
rs763538892 Chr.7: 140734785-140734785 on Build GRCh38
rs775598011 Chr.7: 140734772-140734773 on Build GRCh38

View Full Product Details