Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32350187-32350440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR632
Entrez Gene ID
693217
Gene Name
microRNA 632
Gene Aliases
MIRN632, hsa-mir-632
UniGene
Gene Chromosome Location
Chr.17: 32350109-32350202 on Build GRCh38
Transcripts
NR_030362.1 NM_001032293.2 NM_001098507.1 NM_003457.3 XM_017025019.1 XM_017025020.1

Gene Symbol
ZNF207
Entrez Gene ID
7756
Gene Name
zinc finger protein 207
Gene Aliases
BuGZ, hBuGZ
UniGene
Hs.500775
Gene Chromosome Location
Chr.17: 32350117-32381885 on Build GRCh38
Transcripts
NR_030362.1 NM_001032293.2 NM_001098507.1 NM_003457.3 XM_017025019.1 XM_017025020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6273170 c.41+10G>T p.? Chr.17: 32350336-32350336 on Build GRCh38
4579576 c.10_12delAAG p.K7delK Chr.17: 32350295-32350297 on Build GRCh38
3717279 c.39C>A p.C13* Chr.17: 32350324-32350324 on Build GRCh38
5637223 c.24G>T p.Q8H Chr.17: 32350309-32350309 on Build GRCh38
6273171 c.41+10G>T p.? Chr.17: 32350336-32350336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35933557 Chr.17: 32350392-32350393 on Build GRCh38
rs773544954 Chr.17: 32350209-32350209 on Build GRCh38
rs766585515 Chr.17: 32350257-32350257 on Build GRCh38
rs372983891 Chr.17: 32350241-32350241 on Build GRCh38
rs921055178 Chr.17: 32350236-32350236 on Build GRCh38
rs779597279 Chr.17: 32350275-32350275 on Build GRCh38
rs771194446 Chr.17: 32350327-32350327 on Build GRCh38
rs774251651 Chr.17: 32350347-32350347 on Build GRCh38
rs766082314 Chr.17: 32350377-32350377 on Build GRCh38
rs762737129 Chr.17: 32350227-32350227 on Build GRCh38
rs11538423 Chr.17: 32350271-32350271 on Build GRCh38
rs547463975 Chr.17: 32350414-32350414 on Build GRCh38
rs769471067 Chr.17: 32350216-32350216 on Build GRCh38
rs761907686 Chr.17: 32350223-32350223 on Build GRCh38
rs749135912 Chr.17: 32350336-32350336 on Build GRCh38
rs760447540 Chr.17: 32350247-32350247 on Build GRCh38
rs749415532 Chr.17: 32350272-32350272 on Build GRCh38
rs893352675 Chr.17: 32350331-32350331 on Build GRCh38
rs1010434113 Chr.17: 32350394-32350394 on Build GRCh38
rs562685009 Chr.17: 32350368-32350368 on Build GRCh38
rs758537619 Chr.17: 32350297-32350297 on Build GRCh38
rs202040527 Chr.17: 32350214-32350214 on Build GRCh38
rs1012911894 Chr.17: 32350342-32350342 on Build GRCh38
rs750065952 Chr.17: 32350244-32350244 on Build GRCh38
rs768657070 Chr.17: 32350231-32350231 on Build GRCh38
rs192426798 Chr.17: 32350367-32350367 on Build GRCh38
rs371425544 Chr.17: 32350233-32350233 on Build GRCh38
rs1041379771 Chr.17: 32350399-32350399 on Build GRCh38
rs753304351 Chr.17: 32350285-32350285 on Build GRCh38
rs747132841 Chr.17: 32350310-32350310 on Build GRCh38
rs1003352264 Chr.17: 32350379-32350379 on Build GRCh38
rs1044784363 Chr.17: 32350355-32350355 on Build GRCh38
rs772386223 Chr.17: 32350207-32350207 on Build GRCh38
rs140451700 Chr.17: 32350306-32350306 on Build GRCh38
rs748261615 Chr.17: 32350206-32350206 on Build GRCh38
rs528774844 Chr.17: 32350406-32350406 on Build GRCh38
rs756377321 Chr.17: 32350278-32350279 on Build GRCh38
rs768477954 Chr.17: 32350338-32350338 on Build GRCh38

View Full Product Details