Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24542889-24543158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUCD1
Entrez Gene ID
83606
Gene Name
guanylyl cyclase domain containing 1
Gene Aliases
C22orf13, LLN4
UniGene
Hs.9850
Gene Chromosome Location
Chr.22: 24540423-24555935 on Build GRCh38
Transcripts
NM_001284251.1 NM_001284252.1 NM_001284253.1 NM_001284254.1 NM_001284255.1 NM_001284256.1 NM_001284257.1 NM_031444.3 NR_104286.1 XM_005261761.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3992066 c.703T>A p.F235I Chr.22: 24543026-24543026 on Build GRCh38
4555628 c.661G>C p.E221Q Chr.22: 24543068-24543068 on Build GRCh38
1660882 c.648C>A p.S216R Chr.22: 24543081-24543081 on Build GRCh38
1032724 c.695A>G p.D232G Chr.22: 24543034-24543034 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs377004600 Chr.22: 24542990-24542990 on Build GRCh38
rs746702401 Chr.22: 24543067-24543067 on Build GRCh38
rs199636561 Chr.22: 24543114-24543114 on Build GRCh38
rs925615239 Chr.22: 24542916-24542916 on Build GRCh38
rs757418558 Chr.22: 24543001-24543001 on Build GRCh38
rs370163221 Chr.22: 24543023-24543023 on Build GRCh38
rs748585336 Chr.22: 24543048-24543048 on Build GRCh38
rs780506957 Chr.22: 24543021-24543021 on Build GRCh38
rs749592801 Chr.22: 24543031-24543031 on Build GRCh38
rs367984746 Chr.22: 24543055-24543055 on Build GRCh38
rs749834160 Chr.22: 24542960-24542960 on Build GRCh38
rs573877167 Chr.22: 24542954-24542954 on Build GRCh38
rs757979384 Chr.22: 24543139-24543144 on Build GRCh38
rs115772121 Chr.22: 24542955-24542955 on Build GRCh38
rs960894230 Chr.22: 24543087-24543087 on Build GRCh38
rs144393289 Chr.22: 24543078-24543078 on Build GRCh38
rs201162614 Chr.22: 24542988-24542988 on Build GRCh38
rs762936512 Chr.22: 24543112-24543112 on Build GRCh38
rs945737615 Chr.22: 24543138-24543138 on Build GRCh38
rs769011332 Chr.22: 24542963-24542963 on Build GRCh38
rs200583030 Chr.22: 24542987-24542987 on Build GRCh38
rs779387007 Chr.22: 24543041-24543041 on Build GRCh38
rs755338046 Chr.22: 24543133-24543133 on Build GRCh38
rs761005870 Chr.22: 24543117-24543117 on Build GRCh38
rs750680578 Chr.22: 24543011-24543011 on Build GRCh38
rs765061455 Chr.22: 24543083-24543083 on Build GRCh38
rs889908020 Chr.22: 24542956-24542956 on Build GRCh38
rs766497349 Chr.22: 24543123-24543123 on Build GRCh38
rs8139822 Chr.22: 24542940-24542940 on Build GRCh38
rs765255569 Chr.22: 24542996-24542996 on Build GRCh38
rs755534205 Chr.22: 24543036-24543036 on Build GRCh38
rs747642962 Chr.22: 24542972-24542972 on Build GRCh38
rs377401295 Chr.22: 24542974-24542974 on Build GRCh38
rs780232658 Chr.22: 24542958-24542958 on Build GRCh38
rs767648504 Chr.22: 24543008-24543008 on Build GRCh38
rs771956939 Chr.22: 24543000-24543001 on Build GRCh38
rs1053463962 Chr.22: 24542939-24542939 on Build GRCh38
rs746616470 Chr.22: 24542977-24542977 on Build GRCh38
rs756251318 Chr.22: 24543012-24543012 on Build GRCh38
rs370831364 Chr.22: 24542976-24542976 on Build GRCh38
rs777315979 Chr.22: 24542964-24542964 on Build GRCh38
rs771206421 Chr.22: 24542989-24542989 on Build GRCh38
rs927144177 Chr.22: 24543050-24543050 on Build GRCh38
rs772544173 Chr.22: 24543063-24543063 on Build GRCh38
rs776551490 Chr.22: 24543081-24543081 on Build GRCh38
rs751759544 Chr.22: 24542997-24542997 on Build GRCh38
rs1038300052 Chr.22: 24542966-24542966 on Build GRCh38
rs759249277 Chr.22: 24543082-24543082 on Build GRCh38
rs1005646676 Chr.22: 24542948-24542948 on Build GRCh38
rs775583557 Chr.22: 24543084-24543084 on Build GRCh38
rs935874914 Chr.22: 24542929-24542929 on Build GRCh38
rs374969094 Chr.22: 24543054-24543054 on Build GRCh38
rs979723906 Chr.22: 24542908-24542908 on Build GRCh38
rs202062564 Chr.22: 24543126-24543126 on Build GRCh38

View Full Product Details