Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38300117-38300384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATL2
Entrez Gene ID
64225
Gene Name
atlastin GTPase 2
Gene Aliases
ARL3IP2, ARL6IP2, atlastin2
UniGene
Hs.594950
Gene Chromosome Location
Chr.2: 38293957-38378579 on Build GRCh38
Transcripts
NM_001135673.2 NM_001308076.1 NM_022374.3 NR_024191.1 XM_011533017.1 XM_011533018.2 XM_011533019.2 XM_011533020.1 XM_011533022.2 XM_011533023.2 XM_017004686.1 XM_017004687.1 XM_017004688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4814476 c.1113A>G p.P371P Chr.2: 38300287-38300287 on Build GRCh38
5598032 c.1064C>T p.S355F Chr.2: 38300282-38300282 on Build GRCh38
5598031 c.1118C>T p.S373F Chr.2: 38300282-38300282 on Build GRCh38
3183857 c.1086C>A p.I362I Chr.2: 38300314-38300314 on Build GRCh38
5598030 c.1118C>T p.S373F Chr.2: 38300282-38300282 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945226128 Chr.2: 38300338-38300338 on Build GRCh38
rs866483863 Chr.2: 38300345-38300345 on Build GRCh38
rs369327955 Chr.2: 38300256-38300257 on Build GRCh38
rs749611549 Chr.2: 38300227-38300230 on Build GRCh38
rs376071199 Chr.2: 38300303-38300303 on Build GRCh38
rs986051692 Chr.2: 38300280-38300280 on Build GRCh38
rs374336033 Chr.2: 38300358-38300358 on Build GRCh38
rs764381907 Chr.2: 38300255-38300255 on Build GRCh38
rs537048917 Chr.2: 38300190-38300190 on Build GRCh38
rs77626845 Chr.2: 38300305-38300305 on Build GRCh38
rs775627945 Chr.2: 38300304-38300306 on Build GRCh38
rs761108530 Chr.2: 38300331-38300331 on Build GRCh38
rs997700395 Chr.2: 38300169-38300169 on Build GRCh38
rs761179365 Chr.2: 38300241-38300241 on Build GRCh38
rs777247121 Chr.2: 38300344-38300344 on Build GRCh38
rs762502213 Chr.2: 38300227-38300227 on Build GRCh38
rs879230314 Chr.2: 38300308-38300308 on Build GRCh38
rs964427973 Chr.2: 38300160-38300160 on Build GRCh38
rs962468219 Chr.2: 38300226-38300226 on Build GRCh38
rs749213550 Chr.2: 38300320-38300320 on Build GRCh38
rs750651024 Chr.2: 38300262-38300262 on Build GRCh38
rs762315189 Chr.2: 38300352-38300352 on Build GRCh38
rs751453943 Chr.2: 38300284-38300284 on Build GRCh38
rs765901767 Chr.2: 38300234-38300234 on Build GRCh38
rs41280635 Chr.2: 38300355-38300355 on Build GRCh38
rs145819319 Chr.2: 38300255-38300256 on Build GRCh38
rs982497756 Chr.2: 38300156-38300156 on Build GRCh38
rs368617690 Chr.2: 38300343-38300343 on Build GRCh38
rs754206122 Chr.2: 38300256-38300256 on Build GRCh38
rs569687183 Chr.2: 38300140-38300140 on Build GRCh38
rs201868481 Chr.2: 38300230-38300230 on Build GRCh38
rs758464322 Chr.2: 38300266-38300266 on Build GRCh38
rs748937150 Chr.2: 38300310-38300310 on Build GRCh38
rs765595597 Chr.2: 38300260-38300260 on Build GRCh38
rs369786919 Chr.2: 38300359-38300359 on Build GRCh38
rs371129440 Chr.2: 38300238-38300238 on Build GRCh38
rs984141587 Chr.2: 38300250-38300250 on Build GRCh38
rs917773447 Chr.2: 38300334-38300334 on Build GRCh38
rs777464211 Chr.2: 38300309-38300309 on Build GRCh38
rs796751822 Chr.2: 38300256-38300257 on Build GRCh38
rs1030938335 Chr.2: 38300170-38300170 on Build GRCh38
rs754884875 Chr.2: 38300295-38300295 on Build GRCh38
rs770355830 Chr.2: 38300317-38300317 on Build GRCh38
rs771430470 Chr.2: 38300253-38300254 on Build GRCh38
rs760983151 Chr.2: 38300339-38300341 on Build GRCh38
rs1038692904 Chr.2: 38300360-38300360 on Build GRCh38
rs963352783 Chr.2: 38300264-38300264 on Build GRCh38
rs767607891 Chr.2: 38300256-38300259 on Build GRCh38
rs780176099 Chr.2: 38300269-38300269 on Build GRCh38
rs1015024222 Chr.2: 38300163-38300163 on Build GRCh38

View Full Product Details