Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 107861643-107861904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL17
Entrez Gene ID
64839
Gene Name
F-box and leucine rich repeat protein 17
Gene Aliases
FBXO13, Fbl17, Fbx13
UniGene
Hs.657225
Gene Chromosome Location
Chr.5: 107859033-108382098 on Build GRCh38
Transcripts
NM_001163315.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4759884 c.2045G>T p.R682M Chr.5: 107861781-107861781 on Build GRCh38
5461279 c.2035G>A p.D679N Chr.5: 107861791-107861791 on Build GRCh38
6168576 c.2072G>T p.G691V Chr.5: 107861754-107861754 on Build GRCh38
3607465 c.2081C>T p.P694L Chr.5: 107861745-107861745 on Build GRCh38
3975114 c.2084A>G p.N695S Chr.5: 107861742-107861742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201479758 Chr.5: 107861854-107861854 on Build GRCh38
rs755249881 Chr.5: 107861868-107861868 on Build GRCh38
rs368164862 Chr.5: 107861800-107861800 on Build GRCh38
rs34990078 Chr.5: 107861801-107861801 on Build GRCh38
rs375040831 Chr.5: 107861837-107861837 on Build GRCh38
rs774714019 Chr.5: 107861705-107861705 on Build GRCh38
rs752550566 Chr.5: 107861740-107861740 on Build GRCh38
rs777663695 Chr.5: 107861748-107861748 on Build GRCh38
rs757112502 Chr.5: 107861850-107861850 on Build GRCh38
rs778814643 Chr.5: 107861874-107861874 on Build GRCh38
rs751734539 Chr.5: 107861744-107861744 on Build GRCh38
rs780438502 Chr.5: 107861769-107861769 on Build GRCh38
rs572350610 Chr.5: 107861831-107861831 on Build GRCh38
rs778235887 Chr.5: 107861883-107861883 on Build GRCh38
rs138232838 Chr.5: 107861828-107861828 on Build GRCh38
rs751342494 Chr.5: 107861847-107861847 on Build GRCh38
rs753984329 Chr.5: 107861858-107861858 on Build GRCh38
rs561938338 Chr.5: 107861818-107861818 on Build GRCh38
rs549692367 Chr.5: 107861881-107861881 on Build GRCh38
rs772594905 Chr.5: 107861795-107861795 on Build GRCh38
rs138387486 Chr.5: 107861689-107861689 on Build GRCh38
rs763956267 Chr.5: 107861708-107861708 on Build GRCh38
rs376664117 Chr.5: 107861675-107861675 on Build GRCh38
rs758958421 Chr.5: 107861733-107861733 on Build GRCh38
rs751432009 Chr.5: 107861757-107861757 on Build GRCh38
rs747099433 Chr.5: 107861763-107861763 on Build GRCh38
rs747584091 Chr.5: 107861884-107861884 on Build GRCh38
rs895971089 Chr.5: 107861855-107861855 on Build GRCh38
rs770163016 Chr.5: 107861686-107861686 on Build GRCh38
rs758413862 Chr.5: 107861744-107861744 on Build GRCh38
rs752275308 Chr.5: 107861834-107861834 on Build GRCh38
rs143844606 Chr.5: 107861717-107861717 on Build GRCh38
rs946355528 Chr.5: 107861731-107861731 on Build GRCh38
rs762421958 Chr.5: 107861706-107861706 on Build GRCh38
rs768837022 Chr.5: 107861692-107861692 on Build GRCh38
rs1050647357 Chr.5: 107861668-107861668 on Build GRCh38
rs896163853 Chr.5: 107861691-107861691 on Build GRCh38
rs746067118 Chr.5: 107861678-107861678 on Build GRCh38
rs748259897 Chr.5: 107861877-107861877 on Build GRCh38
rs149163100 Chr.5: 107861846-107861846 on Build GRCh38
rs369567983 Chr.5: 107861718-107861718 on Build GRCh38
rs773495429 Chr.5: 107861798-107861798 on Build GRCh38
rs772403040 Chr.5: 107861707-107861707 on Build GRCh38
rs373905015 Chr.5: 107861772-107861772 on Build GRCh38
rs764870650 Chr.5: 107861738-107861738 on Build GRCh38
rs748553371 Chr.5: 107861784-107861784 on Build GRCh38
rs61749621 Chr.5: 107861719-107861719 on Build GRCh38
rs779099021 Chr.5: 107861773-107861773 on Build GRCh38
rs531559246 Chr.5: 107861825-107861825 on Build GRCh38
rs1043294474 Chr.5: 107861753-107861753 on Build GRCh38

View Full Product Details