Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46354830-46355089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35C2
Entrez Gene ID
51006
Gene Name
solute carrier family 35 member C2
Gene Aliases
BA394O2.1, C20orf5, CGI-15, OVCOV1
UniGene
Hs.593344
Gene Chromosome Location
Chr.20: 46349528-46364461 on Build GRCh38
Transcripts
NM_001281457.1 NM_001281458.1 NM_001281459.1 NM_001281460.1 NM_015945.11 NM_173073.3 NM_173179.3 XM_011528831.1 XM_011528832.1 XM_011528833.1 XM_011528834.1 XM_011528835.1 XM_011528836.1 XM_011528837.1 XM_011528838.1 XM_017027856.1 XM_017027857.1 XM_017027858.1 XM_017027859.1 XM_017027860.1 XM_017027861.1 XM_017027862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7247523 c.262+4C>T p.? Chr.20: 46354878-46354878 on Build GRCh38
4875096 c.232G>A p.G78R Chr.20: 46354912-46354912 on Build GRCh38
7208801 c.586A>T p.N196Y Chr.20: 46354957-46354957 on Build GRCh38
4292652 c.664G>A p.G222S Chr.20: 46355073-46355073 on Build GRCh38
7208802 c.187A>T p.N63Y Chr.20: 46354957-46354957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112467299 Chr.20: 46354967-46354967 on Build GRCh38
rs752180989 Chr.20: 46355068-46355068 on Build GRCh38
rs1040383189 Chr.20: 46354911-46354911 on Build GRCh38
rs768204151 Chr.20: 46354915-46354915 on Build GRCh38
rs745758104 Chr.20: 46354873-46354873 on Build GRCh38
rs775452102 Chr.20: 46355047-46355047 on Build GRCh38
rs140165611 Chr.20: 46355061-46355061 on Build GRCh38
rs776251556 Chr.20: 46354922-46354922 on Build GRCh38
rs773464165 Chr.20: 46354948-46354948 on Build GRCh38
rs781081145 Chr.20: 46354895-46354896 on Build GRCh38
rs749588369 Chr.20: 46355022-46355022 on Build GRCh38
rs770129768 Chr.20: 46354944-46354944 on Build GRCh38
rs760908759 Chr.20: 46355048-46355048 on Build GRCh38
rs576707210 Chr.20: 46355014-46355014 on Build GRCh38
rs752892811 Chr.20: 46354975-46354975 on Build GRCh38
rs151119784 Chr.20: 46355010-46355010 on Build GRCh38
rs1012797679 Chr.20: 46355020-46355020 on Build GRCh38
rs373176914 Chr.20: 46354987-46354987 on Build GRCh38
rs750505883 Chr.20: 46354909-46354910 on Build GRCh38
rs202059286 Chr.20: 46355065-46355065 on Build GRCh38
rs755214455 Chr.20: 46354857-46354857 on Build GRCh38
rs149565718 Chr.20: 46354964-46354964 on Build GRCh38
rs775041650 Chr.20: 46354969-46354969 on Build GRCh38
rs370573129 Chr.20: 46355041-46355041 on Build GRCh38
rs12480667 Chr.20: 46354878-46354878 on Build GRCh38
rs535992725 Chr.20: 46354949-46354949 on Build GRCh38
rs375977599 Chr.20: 46354928-46354928 on Build GRCh38
rs369278101 Chr.20: 46354891-46354891 on Build GRCh38
rs759669175 Chr.20: 46354970-46354970 on Build GRCh38
rs758350844 Chr.20: 46354876-46354876 on Build GRCh38
rs750214603 Chr.20: 46354988-46354988 on Build GRCh38
rs537225524 Chr.20: 46355019-46355019 on Build GRCh38
rs555972774 Chr.20: 46355033-46355033 on Build GRCh38
rs757067779 Chr.20: 46354868-46354868 on Build GRCh38
rs370738121 Chr.20: 46355003-46355003 on Build GRCh38
rs370290945 Chr.20: 46354853-46354853 on Build GRCh38
rs752914736 Chr.20: 46354867-46354867 on Build GRCh38
rs199648810 Chr.20: 46354974-46354974 on Build GRCh38
rs756300684 Chr.20: 46354978-46354978 on Build GRCh38
rs780573785 Chr.20: 46355006-46355006 on Build GRCh38
rs201253430 Chr.20: 46354998-46354998 on Build GRCh38
rs371537544 Chr.20: 46355066-46355066 on Build GRCh38
rs779944841 Chr.20: 46355000-46355000 on Build GRCh38
rs199606072 Chr.20: 46355064-46355064 on Build GRCh38
rs549986139 Chr.20: 46354865-46354865 on Build GRCh38
rs751714801 Chr.20: 46354855-46354855 on Build GRCh38
rs778573323 Chr.20: 46354872-46354872 on Build GRCh38
rs200754558 Chr.20: 46355034-46355034 on Build GRCh38
rs764189719 Chr.20: 46355052-46355052 on Build GRCh38
rs755693821 Chr.20: 46355070-46355070 on Build GRCh38
rs936868976 Chr.20: 46354943-46354943 on Build GRCh38
rs754137411 Chr.20: 46355058-46355058 on Build GRCh38
rs963154012 Chr.20: 46355044-46355044 on Build GRCh38
rs540107970 Chr.20: 46355008-46355008 on Build GRCh38
rs747006886 Chr.20: 46355002-46355002 on Build GRCh38

View Full Product Details