Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157638334-157638573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOX30
Entrez Gene ID
11063
Gene Name
SRY-box 30
Gene Aliases
-
UniGene
Hs.529462
Gene Chromosome Location
Chr.5: 157624796-157672081 on Build GRCh38
Transcripts
NM_001308165.1 NM_007017.2 NM_178424.1 XM_005265803.4 XM_011534420.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4729176 c.1738A>G p.I580V Chr.5: 157638372-157638372 on Build GRCh38
6787491 c.1651A>C p.S551R Chr.5: 157638459-157638459 on Build GRCh38
5434321 c.1567C>T p.Q523* Chr.5: 157638543-157638543 on Build GRCh38
4729177 c.1672G>A p.A558T Chr.5: 157638438-157638438 on Build GRCh38
3429198 c.1727G>A p.R576K Chr.5: 157638383-157638383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145658817 Chr.5: 157638435-157638435 on Build GRCh38
rs200265531 Chr.5: 157638548-157638548 on Build GRCh38
rs748509515 Chr.5: 157638553-157638556 on Build GRCh38
rs754012135 Chr.5: 157638380-157638380 on Build GRCh38
rs563328679 Chr.5: 157638430-157638430 on Build GRCh38
rs112851243 Chr.5: 157638411-157638411 on Build GRCh38
rs150102127 Chr.5: 157638537-157638537 on Build GRCh38
rs142979427 Chr.5: 157638491-157638491 on Build GRCh38
rs760714972 Chr.5: 157638367-157638367 on Build GRCh38
rs75818287 Chr.5: 157638431-157638431 on Build GRCh38
rs373125772 Chr.5: 157638384-157638384 on Build GRCh38
rs138390114 Chr.5: 157638420-157638420 on Build GRCh38
rs754485706 Chr.5: 157638412-157638412 on Build GRCh38
rs138475659 Chr.5: 157638484-157638484 on Build GRCh38
rs762040026 Chr.5: 157638449-157638449 on Build GRCh38
rs765367264 Chr.5: 157638463-157638463 on Build GRCh38
rs750769095 Chr.5: 157638388-157638388 on Build GRCh38
rs758664177 Chr.5: 157638483-157638483 on Build GRCh38
rs755544518 Chr.5: 157638513-157638513 on Build GRCh38
rs192793736 Chr.5: 157638437-157638437 on Build GRCh38
rs776697812 Chr.5: 157638448-157638448 on Build GRCh38
rs941394859 Chr.5: 157638544-157638544 on Build GRCh38
rs145022337 Chr.5: 157638387-157638387 on Build GRCh38
rs757362004 Chr.5: 157638475-157638475 on Build GRCh38
rs35974801 Chr.5: 157638489-157638490 on Build GRCh38
rs780788334 Chr.5: 157638518-157638518 on Build GRCh38
rs750727674 Chr.5: 157638479-157638479 on Build GRCh38
rs565834542 Chr.5: 157638439-157638439 on Build GRCh38
rs752133831 Chr.5: 157638533-157638533 on Build GRCh38
rs533492138 Chr.5: 157638421-157638421 on Build GRCh38
rs370749800 Chr.5: 157638399-157638399 on Build GRCh38
rs113663535 Chr.5: 157638495-157638495 on Build GRCh38
rs749018375 Chr.5: 157638546-157638546 on Build GRCh38
rs149155431 Chr.5: 157638400-157638400 on Build GRCh38
rs764208651 Chr.5: 157638372-157638372 on Build GRCh38
rs755716247 Chr.5: 157638536-157638536 on Build GRCh38
rs73306857 Chr.5: 157638422-157638422 on Build GRCh38

View Full Product Details