Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 108740362-108740617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP2
Entrez Gene ID
5903
Gene Name
RAN binding protein 2
Gene Aliases
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2
UniGene
Hs.199561
Gene Chromosome Location
Chr.2: 108719446-109842301 on Build GRCh38
Transcripts
NM_006267.4 XM_005264002.2 XM_005264003.2 XM_005264004.2 XM_005264005.4 XM_005264007.2 XM_011511575.2 XM_011511576.2 XM_011511578.2 XM_017004623.1 XM_017004624.1 XM_017004625.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6762567 c.820G>A p.G274R Chr.2: 108740526-108740526 on Build GRCh38
5019921 c.816G>C p.L272F Chr.2: 108740522-108740522 on Build GRCh38
4084070 c.829G>A p.E277K Chr.2: 108740535-108740535 on Build GRCh38
1263788 c.894G>A p.L298L Chr.2: 108740600-108740600 on Build GRCh38
4896992 c.812C>T p.S271F Chr.2: 108740518-108740518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772195001 Chr.2: 108740489-108740490 on Build GRCh38
rs572238427 Chr.2: 108740576-108740576 on Build GRCh38
rs199652721 Chr.2: 108740540-108740540 on Build GRCh38
rs995198613 Chr.2: 108740432-108740432 on Build GRCh38
rs781080673 Chr.2: 108740578-108740578 on Build GRCh38
rs375251003 Chr.2: 108740567-108740567 on Build GRCh38
rs369585628 Chr.2: 108740579-108740579 on Build GRCh38
rs748977485 Chr.2: 108740494-108740494 on Build GRCh38
rs2693118 Chr.2: 108740427-108740427 on Build GRCh38
rs758806704 Chr.2: 108740530-108740532 on Build GRCh38
rs796632729 Chr.2: 108740497-108740497 on Build GRCh38
rs779968698 Chr.2: 108740584-108740584 on Build GRCh38
rs531623769 Chr.2: 108740580-108740580 on Build GRCh38
rs761291606 Chr.2: 108740517-108740517 on Build GRCh38
rs778063632 Chr.2: 108740456-108740456 on Build GRCh38
rs771037874 Chr.2: 108740463-108740463 on Build GRCh38
rs537018827 Chr.2: 108740428-108740428 on Build GRCh38
rs76352345 Chr.2: 108740522-108740522 on Build GRCh38
rs765631701 Chr.2: 108740466-108740469 on Build GRCh38
rs149754019 Chr.2: 108740512-108740512 on Build GRCh38
rs1018886885 Chr.2: 108740451-108740451 on Build GRCh38
rs1008868566 Chr.2: 108740448-108740448 on Build GRCh38
rs542732311 Chr.2: 108740577-108740577 on Build GRCh38
rs752998295 Chr.2: 108740560-108740560 on Build GRCh38
rs937017060 Chr.2: 108740559-108740559 on Build GRCh38
rs765638079 Chr.2: 108740555-108740555 on Build GRCh38
rs1028878388 Chr.2: 108740439-108740439 on Build GRCh38
rs773910679 Chr.2: 108740513-108740513 on Build GRCh38
rs372290404 Chr.2: 108740462-108740462 on Build GRCh38
rs745749236 Chr.2: 108740473-108740473 on Build GRCh38
rs138540027 Chr.2: 108740483-108740483 on Build GRCh38
rs556590512 Chr.2: 108740443-108740443 on Build GRCh38
rs750811752 Chr.2: 108740516-108740517 on Build GRCh38
rs781150681 Chr.2: 108740472-108740472 on Build GRCh38
rs553933395 Chr.2: 108740524-108740524 on Build GRCh38
rs543758355 Chr.2: 108740587-108740587 on Build GRCh38
rs953519057 Chr.2: 108740445-108740445 on Build GRCh38
rs374671593 Chr.2: 108740475-108740475 on Build GRCh38

View Full Product Details