Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 107544072-107544339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD11
Entrez Gene ID
121551
Gene Name
BTB domain containing 11
Gene Aliases
ABTB2B
UniGene
Hs.271272
Gene Chromosome Location
Chr.12: 107317168-107659642 on Build GRCh38
Transcripts
NM_001018072.1 XM_005268645.3 XM_011537909.2 XM_011537910.2 XM_017018807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1945364 c.1443G>C p.W481C Chr.12: 107544092-107544092 on Build GRCh38
6071076 c.1479C>A p.L493L Chr.12: 107544128-107544128 on Build GRCh38
5840010 c.1437G>A p.M479I Chr.12: 107544086-107544086 on Build GRCh38
5691175 c.1490G>A p.R497K Chr.12: 107544139-107544139 on Build GRCh38
6071075 c.1479C>A p.L493L Chr.12: 107544128-107544128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952796567 Chr.12: 107544285-107544285 on Build GRCh38
rs543655389 Chr.12: 107544265-107544265 on Build GRCh38
rs200217942 Chr.12: 107544159-107544159 on Build GRCh38
rs1033675397 Chr.12: 107544283-107544283 on Build GRCh38
rs561762599 Chr.12: 107544274-107544274 on Build GRCh38
rs113083485 Chr.12: 107544266-107544266 on Build GRCh38
rs764059161 Chr.12: 107544101-107544101 on Build GRCh38
rs572850345 Chr.12: 107544155-107544155 on Build GRCh38
rs774795055 Chr.12: 107544164-107544165 on Build GRCh38
rs755290917 Chr.12: 107544124-107544124 on Build GRCh38
rs781115874 Chr.12: 107544134-107544134 on Build GRCh38
rs761622643 Chr.12: 107544104-107544104 on Build GRCh38
rs764220543 Chr.12: 107544185-107544186 on Build GRCh38
rs2287159 Chr.12: 107544304-107544304 on Build GRCh38
rs888295153 Chr.12: 107544311-107544311 on Build GRCh38
rs533612203 Chr.12: 107544238-107544238 on Build GRCh38
rs976815765 Chr.12: 107544293-107544293 on Build GRCh38
rs774737041 Chr.12: 107544156-107544156 on Build GRCh38
rs373813559 Chr.12: 107544294-107544294 on Build GRCh38
rs942407220 Chr.12: 107544282-107544282 on Build GRCh38
rs909065596 Chr.12: 107544130-107544130 on Build GRCh38
rs771210663 Chr.12: 107544164-107544164 on Build GRCh38
rs369364661 Chr.12: 107544167-107544167 on Build GRCh38
rs1023530547 Chr.12: 107544102-107544102 on Build GRCh38
rs201206655 Chr.12: 107544163-107544164 on Build GRCh38
rs760210184 Chr.12: 107544188-107544188 on Build GRCh38
rs986374121 Chr.12: 107544260-107544260 on Build GRCh38
rs201824797 Chr.12: 107544183-107544183 on Build GRCh38
rs751738689 Chr.12: 107544117-107544117 on Build GRCh38
rs186417347 Chr.12: 107544202-107544202 on Build GRCh38
rs3217152 Chr.12: 107544146-107544147 on Build GRCh38
rs202103301 Chr.12: 107544308-107544308 on Build GRCh38
rs760584120 Chr.12: 107544094-107544094 on Build GRCh38
rs879116489 Chr.12: 107544168-107544168 on Build GRCh38
rs774832277 Chr.12: 107544185-107544185 on Build GRCh38
rs956999634 Chr.12: 107544181-107544181 on Build GRCh38
rs749830256 Chr.12: 107544147-107544148 on Build GRCh38
rs368855091 Chr.12: 107544105-107544105 on Build GRCh38
rs553325763 Chr.12: 107544303-107544303 on Build GRCh38
rs745495824 Chr.12: 107544171-107544171 on Build GRCh38
rs147374295 Chr.12: 107544103-107544103 on Build GRCh38
rs143266875 Chr.12: 107544159-107544160 on Build GRCh38
rs183173160 Chr.12: 107544147-107544147 on Build GRCh38

View Full Product Details