Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11547939-11548196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLK
Entrez Gene ID
640
Gene Name
BLK proto-oncogene, Src family tyrosine kinase
Gene Aliases
MODY11
UniGene
Hs.146591
Gene Chromosome Location
Chr.8: 11493991-11564599 on Build GRCh38
Transcripts
NM_001715.2 XM_011543824.1 XM_011543825.2 XM_011543827.1 XM_011543828.2 XM_011543829.2 XM_017013730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3778844 c.223C>A p.R75R Chr.8: 11548079-11548079 on Build GRCh38
5835503 c.242_243insG p.E83fs*29 Chr.8: 11548098-11548099 on Build GRCh38
1095173 c.210C>T p.T70T Chr.8: 11548066-11548066 on Build GRCh38
1246132 c.184T>A p.F62I Chr.8: 11548040-11548040 on Build GRCh38
3884972 c.206A>G p.Y69C Chr.8: 11548062-11548062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021728196 Chr.8: 11547972-11547972 on Build GRCh38
rs760550352 Chr.8: 11548147-11548147 on Build GRCh38
rs139297964 Chr.8: 11548066-11548066 on Build GRCh38
rs771224606 Chr.8: 11548075-11548075 on Build GRCh38
rs756421117 Chr.8: 11548162-11548162 on Build GRCh38
rs759343550 Chr.8: 11548170-11548170 on Build GRCh38
rs774171207 Chr.8: 11548082-11548082 on Build GRCh38
rs778435147 Chr.8: 11548108-11548108 on Build GRCh38
rs572862103 Chr.8: 11548081-11548081 on Build GRCh38
rs966281977 Chr.8: 11547980-11547980 on Build GRCh38
rs56185487 Chr.8: 11548114-11548114 on Build GRCh38
rs772755122 Chr.8: 11547989-11547989 on Build GRCh38
rs766373727 Chr.8: 11548163-11548163 on Build GRCh38
rs748864580 Chr.8: 11548059-11548059 on Build GRCh38
rs533147029 Chr.8: 11548168-11548168 on Build GRCh38
rs966249322 Chr.8: 11548153-11548153 on Build GRCh38
rs138972988 Chr.8: 11548043-11548043 on Build GRCh38
rs148704915 Chr.8: 11548087-11548087 on Build GRCh38
rs767324062 Chr.8: 11548085-11548085 on Build GRCh38
rs144839649 Chr.8: 11548138-11548138 on Build GRCh38
rs764767670 Chr.8: 11548163-11548163 on Build GRCh38
rs752301935 Chr.8: 11548166-11548166 on Build GRCh38
rs757834420 Chr.8: 11548167-11548167 on Build GRCh38
rs759190553 Chr.8: 11548159-11548159 on Build GRCh38
rs369967336 Chr.8: 11548143-11548143 on Build GRCh38
rs753710738 Chr.8: 11548162-11548163 on Build GRCh38
rs762522454 Chr.8: 11547974-11547974 on Build GRCh38
rs957601742 Chr.8: 11547969-11547969 on Build GRCh38
rs372649769 Chr.8: 11548170-11548170 on Build GRCh38
rs1040912036 Chr.8: 11548119-11548119 on Build GRCh38
rs989389781 Chr.8: 11547971-11547971 on Build GRCh38
rs374914510 Chr.8: 11548080-11548080 on Build GRCh38
rs765333728 Chr.8: 11548006-11548006 on Build GRCh38
rs1042946025 Chr.8: 11548128-11548128 on Build GRCh38
rs544501735 Chr.8: 11548136-11548136 on Build GRCh38
rs201128403 Chr.8: 11547987-11547987 on Build GRCh38
rs749456754 Chr.8: 11548131-11548131 on Build GRCh38
rs748776290 Chr.8: 11547988-11547988 on Build GRCh38
rs374187873 Chr.8: 11548007-11548007 on Build GRCh38
rs764966833 Chr.8: 11548170-11548172 on Build GRCh38
rs760528244 Chr.8: 11548093-11548093 on Build GRCh38
rs753565455 Chr.8: 11548157-11548157 on Build GRCh38
rs752039663 Chr.8: 11548109-11548109 on Build GRCh38
rs891611801 Chr.8: 11548121-11548121 on Build GRCh38
rs768647234 Chr.8: 11548134-11548134 on Build GRCh38
rs763999345 Chr.8: 11548015-11548015 on Build GRCh38
rs149393791 Chr.8: 11548079-11548079 on Build GRCh38
rs368095361 Chr.8: 11548014-11548014 on Build GRCh38
rs757822963 Chr.8: 11548112-11548112 on Build GRCh38
rs55758736 Chr.8: 11548067-11548067 on Build GRCh38
rs776734677 Chr.8: 11547998-11547998 on Build GRCh38
rs56065800 Chr.8: 11548042-11548042 on Build GRCh38
rs146083915 Chr.8: 11548033-11548033 on Build GRCh38
rs368695092 Chr.8: 11547996-11547996 on Build GRCh38
rs763002345 Chr.8: 11548010-11548010 on Build GRCh38
rs746294075 Chr.8: 11548127-11548127 on Build GRCh38
rs976365880 Chr.8: 11547985-11547985 on Build GRCh38
rs761768063 Chr.8: 11548084-11548084 on Build GRCh38
rs754458822 Chr.8: 11548103-11548103 on Build GRCh38
rs754530814 Chr.8: 11548063-11548063 on Build GRCh38
rs771258207 Chr.8: 11547997-11547997 on Build GRCh38
rs199883114 Chr.8: 11548126-11548126 on Build GRCh38
rs761941756 Chr.8: 11548116-11548117 on Build GRCh38
rs202061540 Chr.8: 11548174-11548174 on Build GRCh38
rs538912310 Chr.8: 11548076-11548076 on Build GRCh38
rs779826827 Chr.8: 11548054-11548054 on Build GRCh38
rs200744308 Chr.8: 11548049-11548049 on Build GRCh38
rs750289049 Chr.8: 11548089-11548089 on Build GRCh38
rs371435863 Chr.8: 11548004-11548004 on Build GRCh38
rs773277622 Chr.8: 11548145-11548145 on Build GRCh38
rs371709219 Chr.8: 11548019-11548019 on Build GRCh38
rs1020461493 Chr.8: 11547978-11547978 on Build GRCh38
rs756487163 Chr.8: 11548171-11548171 on Build GRCh38
rs919928429 Chr.8: 11548074-11548074 on Build GRCh38
rs1010571742 Chr.8: 11548123-11548123 on Build GRCh38
rs372425967 Chr.8: 11548130-11548130 on Build GRCh38
rs560206860 Chr.8: 11547959-11547959 on Build GRCh38
rs751513153 Chr.8: 11548017-11548017 on Build GRCh38
rs369654647 Chr.8: 11548100-11548100 on Build GRCh38
rs755867255 Chr.8: 11548050-11548050 on Build GRCh38
rs746201874 Chr.8: 11548078-11548078 on Build GRCh38
rs766217482 Chr.8: 11548149-11548149 on Build GRCh38
rs774345135 Chr.8: 11548042-11548043 on Build GRCh38
rs759308289 Chr.8: 11547986-11547986 on Build GRCh38

View Full Product Details