Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 69940830-69941077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSN1S1
Entrez Gene ID
1446
Gene Name
casein alpha s1
Gene Aliases
CASA, CSN1
UniGene
Hs.3155
Gene Chromosome Location
Chr.4: 69931062-69946570 on Build GRCh38
Transcripts
NM_001025104.1 NM_001890.1 XM_006714089.2 XM_006714090.2 XM_006714091.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1249318 c.321C>T p.N107N Chr.4: 69941039-69941039 on Build GRCh38
5922384 c.322G>A p.E108K Chr.4: 69941040-69941040 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765495554 Chr.4: 69940931-69940931 on Build GRCh38
rs969201463 Chr.4: 69940989-69940989 on Build GRCh38
rs754371950 Chr.4: 69940983-69940983 on Build GRCh38
rs746830211 Chr.4: 69941017-69941017 on Build GRCh38
rs1027525828 Chr.4: 69940955-69940955 on Build GRCh38
rs934140733 Chr.4: 69940907-69940907 on Build GRCh38
rs72852689 Chr.4: 69940894-69940894 on Build GRCh38
rs565123736 Chr.4: 69940889-69940889 on Build GRCh38
rs745825987 Chr.4: 69941040-69941040 on Build GRCh38
rs1034916118 Chr.4: 69940999-69940999 on Build GRCh38
rs550825724 Chr.4: 69940969-69940969 on Build GRCh38
rs1008546249 Chr.4: 69940913-69940913 on Build GRCh38
rs377090431 Chr.4: 69940898-69940898 on Build GRCh38
rs777469309 Chr.4: 69941011-69941011 on Build GRCh38
rs1005559957 Chr.4: 69941002-69941002 on Build GRCh38
rs190848184 Chr.4: 69940875-69940875 on Build GRCh38
rs932944872 Chr.4: 69940890-69940890 on Build GRCh38
rs995079416 Chr.4: 69940951-69940951 on Build GRCh38
rs1002148953 Chr.4: 69940991-69940991 on Build GRCh38
rs569075084 Chr.4: 69940992-69940992 on Build GRCh38
rs889872429 Chr.4: 69940979-69940979 on Build GRCh38
rs897546836 Chr.4: 69940937-69940937 on Build GRCh38
rs781096882 Chr.4: 69941030-69941030 on Build GRCh38
rs1051279973 Chr.4: 69940893-69940893 on Build GRCh38
rs530076582 Chr.4: 69941007-69941007 on Build GRCh38
rs1008115320 Chr.4: 69940995-69940995 on Build GRCh38
rs778407298 Chr.4: 69941000-69941000 on Build GRCh38
rs548607396 Chr.4: 69941008-69941008 on Build GRCh38
rs766830778 Chr.4: 69941012-69941013 on Build GRCh38
rs770730651 Chr.4: 69941022-69941022 on Build GRCh38

View Full Product Details