Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232456865-232457116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPI
Entrez Gene ID
248
Gene Name
alkaline phosphatase, intestinal
Gene Aliases
IAP
UniGene
Hs.37009
Gene Chromosome Location
Chr.2: 232456112-232460745 on Build GRCh38
Transcripts
NM_001631.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1692165 c.328G>A p.D110N Chr.2: 232456926-232456926 on Build GRCh38
3579082 c.362G>A p.G121E Chr.2: 232456960-232456960 on Build GRCh38
4965099 c.301-4G>A p.? Chr.2: 232456895-232456895 on Build GRCh38
6999431 c.334G>A p.A112T Chr.2: 232456932-232456932 on Build GRCh38
3049243 c.432C>T p.R144R Chr.2: 232457030-232457030 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142255168 Chr.2: 232457023-232457023 on Build GRCh38
rs754395184 Chr.2: 232457084-232457084 on Build GRCh38
rs750421707 Chr.2: 232456911-232456911 on Build GRCh38
rs757332360 Chr.2: 232456945-232456945 on Build GRCh38
rs756226320 Chr.2: 232457070-232457070 on Build GRCh38
rs780627683 Chr.2: 232457005-232457005 on Build GRCh38
rs895533336 Chr.2: 232457092-232457092 on Build GRCh38
rs746380280 Chr.2: 232456952-232456952 on Build GRCh38
rs760292347 Chr.2: 232457028-232457028 on Build GRCh38
rs368674077 Chr.2: 232457067-232457067 on Build GRCh38
rs964737724 Chr.2: 232456991-232456991 on Build GRCh38
rs3111341 Chr.2: 232457036-232457036 on Build GRCh38
rs201841252 Chr.2: 232457004-232457004 on Build GRCh38
rs566677043 Chr.2: 232456886-232456886 on Build GRCh38
rs779375665 Chr.2: 232457021-232457021 on Build GRCh38
rs749694571 Chr.2: 232457013-232457013 on Build GRCh38
rs974771004 Chr.2: 232456930-232456930 on Build GRCh38
rs979679991 Chr.2: 232457034-232457034 on Build GRCh38
rs61732032 Chr.2: 232456931-232456931 on Build GRCh38
rs184809548 Chr.2: 232456894-232456894 on Build GRCh38
rs751723259 Chr.2: 232456934-232456934 on Build GRCh38
rs766628797 Chr.2: 232457092-232457092 on Build GRCh38
rs199922420 Chr.2: 232456891-232456891 on Build GRCh38
rs779532144 Chr.2: 232457088-232457088 on Build GRCh38
rs766407990 Chr.2: 232456980-232456980 on Build GRCh38
rs760074656 Chr.2: 232456977-232456977 on Build GRCh38
rs983346861 Chr.2: 232456896-232456896 on Build GRCh38
rs760613268 Chr.2: 232456919-232456919 on Build GRCh38
rs148744505 Chr.2: 232456961-232456961 on Build GRCh38
rs764944705 Chr.2: 232456986-232456986 on Build GRCh38
rs778025369 Chr.2: 232456933-232456933 on Build GRCh38
rs769179073 Chr.2: 232457014-232457014 on Build GRCh38
rs905553613 Chr.2: 232456998-232456998 on Build GRCh38
rs757418119 Chr.2: 232456993-232456993 on Build GRCh38
rs764394136 Chr.2: 232456957-232456957 on Build GRCh38
rs1026987984 Chr.2: 232457069-232457069 on Build GRCh38
rs751809559 Chr.2: 232456987-232456987 on Build GRCh38
rs761521664 Chr.2: 232456968-232456968 on Build GRCh38
rs781205092 Chr.2: 232456946-232456946 on Build GRCh38
rs772898201 Chr.2: 232456972-232456972 on Build GRCh38
rs371086927 Chr.2: 232456932-232456932 on Build GRCh38
rs374924065 Chr.2: 232457087-232457087 on Build GRCh38
rs781365669 Chr.2: 232457000-232457000 on Build GRCh38
rs143514956 Chr.2: 232457059-232457059 on Build GRCh38
rs770379151 Chr.2: 232456958-232456958 on Build GRCh38
rs371502607 Chr.2: 232457030-232457030 on Build GRCh38
rs541317610 Chr.2: 232456959-232456959 on Build GRCh38
rs758615388 Chr.2: 232456967-232456967 on Build GRCh38
rs146038441 Chr.2: 232456908-232456908 on Build GRCh38
rs762703199 Chr.2: 232457045-232457045 on Build GRCh38
rs768847599 Chr.2: 232456962-232456962 on Build GRCh38
rs976253861 Chr.2: 232456955-232456955 on Build GRCh38
rs368552346 Chr.2: 232457027-232457027 on Build GRCh38
rs774155707 Chr.2: 232456964-232456964 on Build GRCh38
rs774137247 Chr.2: 232456895-232456895 on Build GRCh38
rs148853835 Chr.2: 232457048-232457048 on Build GRCh38
rs983253414 Chr.2: 232457016-232457016 on Build GRCh38
rs770082201 Chr.2: 232456890-232456890 on Build GRCh38
rs759387286 Chr.2: 232456985-232456985 on Build GRCh38
rs766333420 Chr.2: 232457071-232457071 on Build GRCh38
rs761424272 Chr.2: 232456906-232456906 on Build GRCh38
rs377393587 Chr.2: 232456892-232456892 on Build GRCh38
rs765035413 Chr.2: 232457031-232457031 on Build GRCh38
rs750554208 Chr.2: 232457001-232457001 on Build GRCh38
rs753704888 Chr.2: 232456983-232456983 on Build GRCh38
rs771730709 Chr.2: 232456969-232456969 on Build GRCh38
rs7559279 Chr.2: 232457029-232457029 on Build GRCh38
rs767790837 Chr.2: 232457058-232457058 on Build GRCh38

View Full Product Details