Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145918811-145918544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX11B
Entrez Gene ID
8799
Gene Name
peroxisomal biogenesis factor 11 beta
Gene Aliases
PEX11-BETA, PEX14B
UniGene
Hs.714608
Gene Chromosome Location
Chr.1: 145911348-145918924 on Build GRCh38
Transcripts
NM_003846.2 NR_073491.1 NR_073492.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7383613 c.8C>T p.A3V Chr.1: 145918681-145918681 on Build GRCh38

View Full Product Details