Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42960669-42960916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNPY3-GNMT
Entrez Gene ID
107080644
Gene Name
CNPY3-GNMT readthrough
Gene Aliases
-
UniGene
Hs.144914
Gene Chromosome Location
Chr.6: 42929130-42963880 on Build GRCh38
Transcripts
NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1 NM_001318865.1 NM_018960.5 NR_134899.1

Gene Symbol
GNMT
Entrez Gene ID
27232
Gene Name
glycine N-methyltransferase
Gene Aliases
HEL-S-182mP
UniGene
Hs.144914
Gene Chromosome Location
Chr.6: 42960754-42963880 on Build GRCh38
Transcripts
NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1 NM_001318865.1 NM_018960.5 NR_134899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6433305 c.16T>C p.Y6H Chr.6: 42960783-42960783 on Build GRCh38
5474947 c.116G>A p.R39H Chr.6: 42960883-42960883 on Build GRCh38
7426764 c.62A>G p.Q21R Chr.6: 42960829-42960829 on Build GRCh38
6047530 c.106G>A p.G36R Chr.6: 42960873-42960873 on Build GRCh38
6388723 c.128C>A p.A43D Chr.6: 42960895-42960895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547669927 Chr.6: 42960706-42960706 on Build GRCh38
rs990336887 Chr.6: 42960829-42960829 on Build GRCh38
rs752961004 Chr.6: 42960816-42960816 on Build GRCh38
rs538620810 Chr.6: 42960855-42960855 on Build GRCh38
rs370044528 Chr.6: 42960779-42960779 on Build GRCh38
rs939023329 Chr.6: 42960881-42960881 on Build GRCh38
rs764250110 Chr.6: 42960834-42960834 on Build GRCh38
rs527633313 Chr.6: 42960705-42960705 on Build GRCh38
rs755908708 Chr.6: 42960886-42960886 on Build GRCh38
rs888403943 Chr.6: 42960878-42960878 on Build GRCh38
rs906041125 Chr.6: 42960717-42960717 on Build GRCh38
rs10948059 Chr.6: 42960723-42960723 on Build GRCh38
rs751634549 Chr.6: 42960835-42960835 on Build GRCh38
rs745619141 Chr.6: 42960880-42960880 on Build GRCh38
rs768138951 Chr.6: 42960766-42960766 on Build GRCh38
rs1045997427 Chr.6: 42960850-42960850 on Build GRCh38
rs766825740 Chr.6: 42960793-42960793 on Build GRCh38
rs376137577 Chr.6: 42960738-42960738 on Build GRCh38
rs975264935 Chr.6: 42960782-42960782 on Build GRCh38
rs777001898 Chr.6: 42960801-42960801 on Build GRCh38
rs112336061 Chr.6: 42960854-42960854 on Build GRCh38
rs35469448 Chr.6: 42960795-42960796 on Build GRCh38
rs200161633 Chr.6: 42960747-42960747 on Build GRCh38
rs1018922856 Chr.6: 42960772-42960772 on Build GRCh38
rs773780925 Chr.6: 42960777-42960777 on Build GRCh38
rs922049356 Chr.6: 42960807-42960807 on Build GRCh38
rs894853034 Chr.6: 42960715-42960715 on Build GRCh38
rs946310687 Chr.6: 42960849-42960849 on Build GRCh38
rs913376414 Chr.6: 42960843-42960843 on Build GRCh38
rs781013631 Chr.6: 42960872-42960872 on Build GRCh38
rs759988463 Chr.6: 42960808-42960808 on Build GRCh38
rs765485319 Chr.6: 42960813-42960813 on Build GRCh38
rs1025498839 Chr.6: 42960819-42960819 on Build GRCh38
rs769388348 Chr.6: 42960745-42960745 on Build GRCh38
rs758753432 Chr.6: 42960821-42960821 on Build GRCh38
rs866499041 Chr.6: 42960823-42960823 on Build GRCh38
rs1004741384 Chr.6: 42960748-42960748 on Build GRCh38
rs762385341 Chr.6: 42960762-42960762 on Build GRCh38

View Full Product Details