Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92342845-92343086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKIB1
Entrez Gene ID
54467
Gene Name
ankyrin repeat and IBR domain containing 1
Gene Aliases
-
UniGene
Hs.83293
Gene Chromosome Location
Chr.7: 92246216-92401773 on Build GRCh38
Transcripts
NM_019004.1 XM_005250458.3 XM_011516334.1 XM_011516335.2 XM_017012359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3883357 c.836A>G p.N279S Chr.7: 92343072-92343072 on Build GRCh38
7284577 c.848G>A p.R283Q Chr.7: 92343084-92343084 on Build GRCh38
3642255 c.824C>T p.S275F Chr.7: 92343060-92343060 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775728635 Chr.7: 92342975-92342975 on Build GRCh38
rs764411428 Chr.7: 92343006-92343006 on Build GRCh38
rs754157093 Chr.7: 92343024-92343024 on Build GRCh38
rs758831784 Chr.7: 92343067-92343067 on Build GRCh38
rs967206644 Chr.7: 92342968-92342968 on Build GRCh38
rs375036574 Chr.7: 92342886-92342886 on Build GRCh38
rs765630513 Chr.7: 92342936-92342936 on Build GRCh38
rs765539705 Chr.7: 92343063-92343063 on Build GRCh38
rs545870700 Chr.7: 92342896-92342896 on Build GRCh38
rs747218018 Chr.7: 92342944-92342944 on Build GRCh38
rs762211381 Chr.7: 92343060-92343060 on Build GRCh38
rs1002270744 Chr.7: 92342880-92342880 on Build GRCh38
rs760898191 Chr.7: 92342983-92342983 on Build GRCh38
rs1021023265 Chr.7: 92342958-92342958 on Build GRCh38
rs768661776 Chr.7: 92343056-92343056 on Build GRCh38
rs750899321 Chr.7: 92343064-92343064 on Build GRCh38

View Full Product Details