Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 36171123-36171348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCIN
Entrez Gene ID
881
Gene Name
calicin
Gene Aliases
BTBD20, KBTBD14
UniGene
Hs.115460
Gene Chromosome Location
Chr.9: 36169392-36171334 on Build GRCh38
Transcripts
NM_005893.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7246939 c.1746C>A p.C582* Chr.9: 36171248-36171248 on Build GRCh38
1108893 c.1660G>T p.D554Y Chr.9: 36171162-36171162 on Build GRCh38
3657335 c.1742C>T p.P581L Chr.9: 36171244-36171244 on Build GRCh38
3952706 c.1746C>T p.C582C Chr.9: 36171248-36171248 on Build GRCh38
159819 c.1693C>G p.P565A Chr.9: 36171195-36171195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570522561 Chr.9: 36171228-36171228 on Build GRCh38
rs778072508 Chr.9: 36171267-36171267 on Build GRCh38
rs746703060 Chr.9: 36171300-36171300 on Build GRCh38
rs772976897 Chr.9: 36171318-36171318 on Build GRCh38
rs75257915 Chr.9: 36171245-36171245 on Build GRCh38
rs761796443 Chr.9: 36171152-36171152 on Build GRCh38
rs113222879 Chr.9: 36171200-36171200 on Build GRCh38
rs766649225 Chr.9: 36171195-36171195 on Build GRCh38
rs1007336903 Chr.9: 36171283-36171283 on Build GRCh38
rs773347840 Chr.9: 36171186-36171186 on Build GRCh38
rs763128938 Chr.9: 36171321-36171321 on Build GRCh38
rs923328445 Chr.9: 36171306-36171306 on Build GRCh38
rs777564675 Chr.9: 36171219-36171219 on Build GRCh38
rs186367350 Chr.9: 36171278-36171278 on Build GRCh38
rs376888107 Chr.9: 36171291-36171291 on Build GRCh38
rs191171185 Chr.9: 36171311-36171311 on Build GRCh38
rs370261629 Chr.9: 36171310-36171310 on Build GRCh38
rs747873387 Chr.9: 36171313-36171313 on Build GRCh38
rs756749264 Chr.9: 36171258-36171258 on Build GRCh38
rs751687018 Chr.9: 36171234-36171234 on Build GRCh38
rs756009275 Chr.9: 36171192-36171192 on Build GRCh38
rs78745631 Chr.9: 36171317-36171317 on Build GRCh38
rs769719968 Chr.9: 36171159-36171159 on Build GRCh38
rs749962330 Chr.9: 36171271-36171271 on Build GRCh38
rs141946752 Chr.9: 36171203-36171203 on Build GRCh38
rs952652922 Chr.9: 36171198-36171198 on Build GRCh38

View Full Product Details