Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 108729029-108729268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP2
Entrez Gene ID
5903
Gene Name
RAN binding protein 2
Gene Aliases
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2
UniGene
Hs.199561
Gene Chromosome Location
Chr.2: 108719446-109842301 on Build GRCh38
Transcripts
NM_006267.4 XM_005264002.2 XM_005264003.2 XM_005264004.2 XM_005264005.4 XM_005264007.2 XM_011511575.2 XM_011511576.2 XM_011511578.2 XM_017004623.1 XM_017004624.1 XM_017004625.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1005132 c.136A>G p.K46E Chr.2: 108729195-108729195 on Build GRCh38
3565101 c.81G>A p.M27I Chr.2: 108729140-108729140 on Build GRCh38
5438439 c.77C>T p.S26L Chr.2: 108729136-108729136 on Build GRCh38
4839114 c.81G>T p.M27I Chr.2: 108729140-108729140 on Build GRCh38
1641610 c.73-10delT p.? Chr.2: 108729122-108729122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758608966 Chr.2: 108729216-108729216 on Build GRCh38
rs536459426 Chr.2: 108729121-108729121 on Build GRCh38
rs766909917 Chr.2: 108729202-108729202 on Build GRCh38
rs759386820 Chr.2: 108729099-108729099 on Build GRCh38
rs747129006 Chr.2: 108729218-108729218 on Build GRCh38
rs537134678 Chr.2: 108729217-108729217 on Build GRCh38
rs753101262 Chr.2: 108729215-108729215 on Build GRCh38
rs763793018 Chr.2: 108729183-108729183 on Build GRCh38
rs773232332 Chr.2: 108729150-108729150 on Build GRCh38
rs747035475 Chr.2: 108729233-108729233 on Build GRCh38
rs762793651 Chr.2: 108729228-108729228 on Build GRCh38
rs755428275 Chr.2: 108729208-108729208 on Build GRCh38
rs576251747 Chr.2: 108729132-108729132 on Build GRCh38
rs775210764 Chr.2: 108729226-108729226 on Build GRCh38
rs75447096 Chr.2: 108729130-108729130 on Build GRCh38
rs764626030 Chr.2: 108729116-108729117 on Build GRCh38
rs756735629 Chr.2: 108729199-108729199 on Build GRCh38
rs780522067 Chr.2: 108729119-108729119 on Build GRCh38
rs761413396 Chr.2: 108729234-108729234 on Build GRCh38
rs758615393 Chr.2: 108729223-108729223 on Build GRCh38
rs771041997 Chr.2: 108729161-108729161 on Build GRCh38
rs1035687684 Chr.2: 108729174-108729174 on Build GRCh38
rs765026716 Chr.2: 108729105-108729105 on Build GRCh38
rs757595735 Chr.2: 108729218-108729219 on Build GRCh38
rs185251250 Chr.2: 108729079-108729079 on Build GRCh38
rs183830746 Chr.2: 108729233-108729233 on Build GRCh38
rs779150518 Chr.2: 108729212-108729212 on Build GRCh38
rs374997577 Chr.2: 108729151-108729151 on Build GRCh38
rs748340440 Chr.2: 108729137-108729137 on Build GRCh38
rs781376773 Chr.2: 108729222-108729222 on Build GRCh38
rs751086162 Chr.2: 108729187-108729187 on Build GRCh38
rs745817784 Chr.2: 108729223-108729223 on Build GRCh38
rs192504405 Chr.2: 108729086-108729086 on Build GRCh38
rs776682099 Chr.2: 108729162-108729162 on Build GRCh38
rs763521631 Chr.2: 108729111-108729111 on Build GRCh38
rs765149627 Chr.2: 108729170-108729170 on Build GRCh38
rs762512927 Chr.2: 108729180-108729180 on Build GRCh38
rs769783915 Chr.2: 108729225-108729225 on Build GRCh38
rs369816017 Chr.2: 108729222-108729223 on Build GRCh38
rs771168683 Chr.2: 108729220-108729220 on Build GRCh38
rs775300579 Chr.2: 108729177-108729177 on Build GRCh38
rs758023257 Chr.2: 108729106-108729106 on Build GRCh38
rs201485597 Chr.2: 108729126-108729126 on Build GRCh38
rs780093909 Chr.2: 108729224-108729225 on Build GRCh38
rs759509405 Chr.2: 108729166-108729166 on Build GRCh38
rs890344821 Chr.2: 108729054-108729054 on Build GRCh38
rs907518574 Chr.2: 108729209-108729209 on Build GRCh38
rs779208982 Chr.2: 108729136-108729136 on Build GRCh38
rs768240263 Chr.2: 108729232-108729232 on Build GRCh38
rs751011002 Chr.2: 108729115-108729115 on Build GRCh38
rs779199214 Chr.2: 108729219-108729219 on Build GRCh38
rs999529430 Chr.2: 108729074-108729074 on Build GRCh38
rs772271582 Chr.2: 108729138-108729138 on Build GRCh38
rs776627359 Chr.2: 108729097-108729097 on Build GRCh38
rs754430705 Chr.2: 108729122-108729122 on Build GRCh38
rs756678963 Chr.2: 108729116-108729116 on Build GRCh38
rs754373974 Chr.2: 108729207-108729207 on Build GRCh38

View Full Product Details