Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 88955106-88955345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KYAT3
Entrez Gene ID
56267
Gene Name
kynurenine aminotransferase 3
Gene Aliases
CCBL2, KAT3, KATIII
UniGene
Hs.481898
Gene Chromosome Location
Chr.1: 88935773-88992960 on Build GRCh38
Transcripts
NM_001008661.2 NM_001008662.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3493130 c.814A>G p.T272A Chr.1: 88955199-88955199 on Build GRCh38
6271806 c.719C>A p.T240K Chr.1: 88955192-88955192 on Build GRCh38
3493131 c.712A>G p.T238A Chr.1: 88955199-88955199 on Build GRCh38
5520082 c.715A>C p.I239L Chr.1: 88955196-88955196 on Build GRCh38
5847436 c.796C>T p.P266S Chr.1: 88955217-88955217 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12095620 Chr.1: 88955142-88955142 on Build GRCh38
rs1033414875 Chr.1: 88955287-88955287 on Build GRCh38
rs781055491 Chr.1: 88955147-88955147 on Build GRCh38
rs760959046 Chr.1: 88955270-88955270 on Build GRCh38
rs780930496 Chr.1: 88955235-88955235 on Build GRCh38
rs772126036 Chr.1: 88955178-88955178 on Build GRCh38
rs769758817 Chr.1: 88955166-88955166 on Build GRCh38
rs149546056 Chr.1: 88955224-88955224 on Build GRCh38
rs757985849 Chr.1: 88955140-88955140 on Build GRCh38
rs745728650 Chr.1: 88955245-88955245 on Build GRCh38
rs769705868 Chr.1: 88955246-88955246 on Build GRCh38
rs754365025 Chr.1: 88955294-88955294 on Build GRCh38
rs770741752 Chr.1: 88955144-88955144 on Build GRCh38
rs35387101 Chr.1: 88955297-88955297 on Build GRCh38
rs771319653 Chr.1: 88955276-88955276 on Build GRCh38
rs777248500 Chr.1: 88955217-88955217 on Build GRCh38
rs189786376 Chr.1: 88955263-88955263 on Build GRCh38
rs1054295603 Chr.1: 88955302-88955302 on Build GRCh38
rs760909604 Chr.1: 88955187-88955187 on Build GRCh38
rs899051483 Chr.1: 88955237-88955237 on Build GRCh38
rs376708393 Chr.1: 88955179-88955179 on Build GRCh38
rs773260426 Chr.1: 88955265-88955265 on Build GRCh38
rs993828211 Chr.1: 88955238-88955238 on Build GRCh38
rs540876914 Chr.1: 88955195-88955195 on Build GRCh38
rs780052439 Chr.1: 88955249-88955249 on Build GRCh38
rs766873510 Chr.1: 88955189-88955189 on Build GRCh38
rs745779521 Chr.1: 88955157-88955157 on Build GRCh38
rs145664845 Chr.1: 88955210-88955210 on Build GRCh38
rs188058829 Chr.1: 88955168-88955168 on Build GRCh38
rs760092730 Chr.1: 88955206-88955206 on Build GRCh38
rs866535900 Chr.1: 88955152-88955152 on Build GRCh38
rs373043582 Chr.1: 88955196-88955196 on Build GRCh38
rs777542818 Chr.1: 88955141-88955141 on Build GRCh38
rs1055518734 Chr.1: 88955151-88955151 on Build GRCh38
rs751243286 Chr.1: 88955222-88955222 on Build GRCh38
rs762009387 Chr.1: 88955174-88955174 on Build GRCh38
rs201465072 Chr.1: 88955213-88955213 on Build GRCh38
rs1000557682 Chr.1: 88955282-88955282 on Build GRCh38
rs903088577 Chr.1: 88955281-88955281 on Build GRCh38
rs369868621 Chr.1: 88955255-88955255 on Build GRCh38

View Full Product Details