Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60902269-60902508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC8
Entrez Gene ID
1161
Gene Name
ERCC excision repair 8, CSA ubiquitin ligase complex subunit
Gene Aliases
CKN1, CSA, UVSS2
UniGene
Hs.435237
Gene Chromosome Location
Chr.5: 60873832-60945078 on Build GRCh38
Transcripts
NM_000082.3 NM_001007233.2 NM_001290285.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3947638 c.581G>T p.W194L Chr.5: 60902478-60902478 on Build GRCh38
3947637 c.582G>T p.W194C Chr.5: 60902477-60902477 on Build GRCh38
6227242 c.617+5A>T p.? Chr.5: 60902437-60902437 on Build GRCh38
5061138 c.604T>G p.L202V Chr.5: 60902455-60902455 on Build GRCh38
3855792 c.554A>G p.H185R Chr.5: 60902505-60902505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771793120 Chr.5: 60902423-60902423 on Build GRCh38
rs763453717 Chr.5: 60902469-60902469 on Build GRCh38
rs755891113 Chr.5: 60902424-60902424 on Build GRCh38
rs747208043 Chr.5: 60902435-60902435 on Build GRCh38
rs750051495 Chr.5: 60902394-60902394 on Build GRCh38
rs551788943 Chr.5: 60902333-60902333 on Build GRCh38
rs930180817 Chr.5: 60902306-60902306 on Build GRCh38
rs121434326 Chr.5: 60902446-60902446 on Build GRCh38
rs777163867 Chr.5: 60902397-60902397 on Build GRCh38
rs76157730 Chr.5: 60902470-60902470 on Build GRCh38
rs764483506 Chr.5: 60902393-60902393 on Build GRCh38
rs536463212 Chr.5: 60902426-60902426 on Build GRCh38
rs534264957 Chr.5: 60902390-60902390 on Build GRCh38
rs765986851 Chr.5: 60902478-60902478 on Build GRCh38
rs760389824 Chr.5: 60902475-60902475 on Build GRCh38
rs281875223 Chr.5: 60902477-60902477 on Build GRCh38
rs571540096 Chr.5: 60902336-60902336 on Build GRCh38
rs4647105 Chr.5: 60902460-60902460 on Build GRCh38
rs898129938 Chr.5: 60902376-60902376 on Build GRCh38
rs201579764 Chr.5: 60902400-60902400 on Build GRCh38
rs1035630201 Chr.5: 60902463-60902463 on Build GRCh38
rs1001955880 Chr.5: 60902380-60902380 on Build GRCh38
rs376937131 Chr.5: 60902415-60902415 on Build GRCh38
rs771262517 Chr.5: 60902436-60902436 on Build GRCh38
rs754949729 Chr.5: 60902408-60902408 on Build GRCh38
rs770467685 Chr.5: 60902451-60902451 on Build GRCh38
rs1016643914 Chr.5: 60902343-60902346 on Build GRCh38
rs1033405110 Chr.5: 60902397-60902399 on Build GRCh38
rs281875224 Chr.5: 60902454-60902454 on Build GRCh38
rs755736782 Chr.5: 60902395-60902395 on Build GRCh38
rs751942615 Chr.5: 60902472-60902472 on Build GRCh38
rs886197550 Chr.5: 60902382-60902382 on Build GRCh38

View Full Product Details