Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32207204-32207475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCC
Entrez Gene ID
55721
Gene Name
IQ motif containing C
Gene Aliases
-
UniGene
Hs.274356
Gene Chromosome Location
Chr.1: 32205635-32208687 on Build GRCh38
Transcripts
NM_001160042.1 NM_018134.2 XM_011541795.2 XM_017001814.1 XM_017001815.1 XM_017001817.1 XM_017001818.1 XM_017001819.1 XM_017001821.1 XM_017001827.1 XM_017001828.1

Gene Symbol
TMEM234
Entrez Gene ID
56063
Gene Name
transmembrane protein 234
Gene Aliases
AASL548, C1orf91, PRO1105, RP4-622L5, dJ622L5.7
UniGene
Hs.272299
Gene Chromosome Location
Chr.1: 32204807-32222371 on Build GRCh38
Transcripts
NM_001160042.1 NM_018134.2 XM_011541795.2 XM_017001814.1 XM_017001815.1 XM_017001817.1 XM_017001818.1 XM_017001819.1 XM_017001821.1 XM_017001827.1 XM_017001828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3689605 c.622G>A p.V208M Chr.1: 32207303-32207303 on Build GRCh38
3488106 c.878G>A p.G293E Chr.1: 32207319-32207319 on Build GRCh38
7214932 c.866T>G p.F289C Chr.1: 32207307-32207307 on Build GRCh38
6576730 c.612G>C p.E204D Chr.1: 32207293-32207293 on Build GRCh38
3789885 c.958G>C p.E320Q Chr.1: 32207399-32207399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765220511 Chr.1: 32207236-32207236 on Build GRCh38
rs113492537 Chr.1: 32207365-32207365 on Build GRCh38
rs769530176 Chr.1: 32207296-32207296 on Build GRCh38
rs758923443 Chr.1: 32207328-32207328 on Build GRCh38
rs747798527 Chr.1: 32207356-32207356 on Build GRCh38
rs3903683 Chr.1: 32207307-32207307 on Build GRCh38
rs754164233 Chr.1: 32207260-32207260 on Build GRCh38
rs754833868 Chr.1: 32207397-32207397 on Build GRCh38
rs767627228 Chr.1: 32207241-32207241 on Build GRCh38
rs375598262 Chr.1: 32207442-32207442 on Build GRCh38
rs776149067 Chr.1: 32207287-32207287 on Build GRCh38
rs749095523 Chr.1: 32207358-32207358 on Build GRCh38
rs540852098 Chr.1: 32207403-32207403 on Build GRCh38
rs767295632 Chr.1: 32207436-32207436 on Build GRCh38
rs367708853 Chr.1: 32207292-32207292 on Build GRCh38
rs902496027 Chr.1: 32207361-32207361 on Build GRCh38
rs890548236 Chr.1: 32207316-32207316 on Build GRCh38
rs1031989779 Chr.1: 32207441-32207441 on Build GRCh38
rs768665888 Chr.1: 32207380-32207380 on Build GRCh38
rs750756942 Chr.1: 32207393-32207393 on Build GRCh38
rs764236869 Chr.1: 32207243-32207243 on Build GRCh38
rs765735506 Chr.1: 32207308-32207308 on Build GRCh38
rs770788837 Chr.1: 32207363-32207363 on Build GRCh38
rs150393107 Chr.1: 32207357-32207357 on Build GRCh38
rs369321515 Chr.1: 32207344-32207344 on Build GRCh38
rs371494687 Chr.1: 32207298-32207298 on Build GRCh38
rs200109284 Chr.1: 32207300-32207300 on Build GRCh38
rs559539057 Chr.1: 32207414-32207414 on Build GRCh38
rs750846956 Chr.1: 32207311-32207311 on Build GRCh38
rs564797132 Chr.1: 32207337-32207337 on Build GRCh38
rs775492029 Chr.1: 32207301-32207301 on Build GRCh38
rs963280771 Chr.1: 32207376-32207376 on Build GRCh38
rs760796095 Chr.1: 32207379-32207379 on Build GRCh38
rs778645525 Chr.1: 32207246-32207248 on Build GRCh38
rs759592940 Chr.1: 32207240-32207240 on Build GRCh38
rs374945736 Chr.1: 32207302-32207302 on Build GRCh38
rs773194857 Chr.1: 32207238-32207238 on Build GRCh38
rs201910762 Chr.1: 32207278-32207278 on Build GRCh38
rs780531095 Chr.1: 32207417-32207417 on Build GRCh38
rs752807316 Chr.1: 32207242-32207242 on Build GRCh38
rs765577015 Chr.1: 32207389-32207389 on Build GRCh38
rs553568223 Chr.1: 32207283-32207283 on Build GRCh38
rs774339310 Chr.1: 32207372-32207372 on Build GRCh38
rs200682317 Chr.1: 32207279-32207279 on Build GRCh38
rs201818705 Chr.1: 32207234-32207234 on Build GRCh38
rs779461068 Chr.1: 32207277-32207277 on Build GRCh38
rs12127777 Chr.1: 32207384-32207384 on Build GRCh38
rs762387921 Chr.1: 32207317-32207318 on Build GRCh38
rs761899517 Chr.1: 32207386-32207386 on Build GRCh38
rs761523250 Chr.1: 32207233-32207233 on Build GRCh38
rs770573300 Chr.1: 32207441-32207441 on Build GRCh38
rs747000628 Chr.1: 32207247-32207253 on Build GRCh38
rs752060024 Chr.1: 32207405-32207405 on Build GRCh38
rs143971469 Chr.1: 32207284-32207284 on Build GRCh38
rs12032332 Chr.1: 32207331-32207331 on Build GRCh38
rs145580199 Chr.1: 32207303-32207303 on Build GRCh38
rs199905667 Chr.1: 32207265-32207265 on Build GRCh38

View Full Product Details