Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145917961-145917688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX11B
Entrez Gene ID
8799
Gene Name
peroxisomal biogenesis factor 11 beta
Gene Aliases
PEX11-BETA, PEX14B
UniGene
Hs.714608
Gene Chromosome Location
Chr.1: 145911348-145918924 on Build GRCh38
Transcripts
NM_001184795.1 NM_003846.2 NR_073491.1 NR_073492.1 NR_073493.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6744088 c.161T>C p.L54P Chr.1: 145917712-145917712 on Build GRCh38
1688457 c.160C>T p.L54F Chr.1: 145917713-145917713 on Build GRCh38
6744085 c.103C>T p.H35Y Chr.1: 145917770-145917770 on Build GRCh38
5341516 c.137G>A p.R46Q Chr.1: 145917736-145917736 on Build GRCh38
1637006 c.111C>T p.A37A Chr.1: 145917762-145917762 on Build GRCh38
View More

View Full Product Details