Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50571022-50571265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHKB-CPT1B
Entrez Gene ID
386593
Gene Name
CHKB-CPT1B readthrough (NMD candidate)
Gene Aliases
CHKL-CPT1B
UniGene
Hs.439777
Gene Chromosome Location
Chr.22: 50568861-50582999 on Build GRCh38
Transcripts
NR_027928.2 NM_001145134.1 NM_001145135.1 NM_001145137.1 NM_004377.3 NM_152245.2 NM_152246.2

Gene Symbol
CPT1B
Entrez Gene ID
1375
Gene Name
carnitine palmitoyltransferase 1B
Gene Aliases
CPT1-M, CPT1M, CPTI, CPTI-M, M-CPT1, MCCPT1, MCPT1
UniGene
Hs.439777
Gene Chromosome Location
Chr.22: 50568861-50578667 on Build GRCh38
Transcripts
NR_027928.2 NM_001145134.1 NM_001145135.1 NM_001145137.1 NM_004377.3 NM_152245.2 NM_152246.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7212387 c.1857G>A p.M619I Chr.22: 50571176-50571176 on Build GRCh38
1417171 c.1793G>A p.R598Q Chr.22: 50571240-50571240 on Build GRCh38
445188 c.1783A>G p.R595G Chr.22: 50571250-50571250 on Build GRCh38
5514370 c.1876-4G>A p.? Chr.22: 50571047-50571047 on Build GRCh38
5859718 c.1864G>T p.G622W Chr.22: 50571169-50571169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs41309651 Chr.22: 50571058-50571058 on Build GRCh38
rs1015594627 Chr.22: 50571183-50571183 on Build GRCh38
rs781499627 Chr.22: 50571212-50571212 on Build GRCh38
rs778963185 Chr.22: 50571043-50571060 on Build GRCh38
rs1030935274 Chr.22: 50571096-50571096 on Build GRCh38
rs548589066 Chr.22: 50571090-50571090 on Build GRCh38
rs111375437 Chr.22: 50571042-50571042 on Build GRCh38
rs763642100 Chr.22: 50571120-50571120 on Build GRCh38
rs571519095 Chr.22: 50571216-50571216 on Build GRCh38
rs763034084 Chr.22: 50571050-50571050 on Build GRCh38
rs141885850 Chr.22: 50571231-50571231 on Build GRCh38
rs776825267 Chr.22: 50571048-50571048 on Build GRCh38
rs759309831 Chr.22: 50571081-50571081 on Build GRCh38
rs373274591 Chr.22: 50571139-50571139 on Build GRCh38
rs147955265 Chr.22: 50571226-50571226 on Build GRCh38
rs745866439 Chr.22: 50571240-50571240 on Build GRCh38
rs750905232 Chr.22: 50571200-50571200 on Build GRCh38
rs370319926 Chr.22: 50571184-50571184 on Build GRCh38
rs753347209 Chr.22: 50571193-50571193 on Build GRCh38
rs563717431 Chr.22: 50571165-50571165 on Build GRCh38
rs747299769 Chr.22: 50571148-50571148 on Build GRCh38
rs9628215 Chr.22: 50571057-50571057 on Build GRCh38
rs997901036 Chr.22: 50571089-50571089 on Build GRCh38
rs763727956 Chr.22: 50571185-50571185 on Build GRCh38
rs770998857 Chr.22: 50571162-50571162 on Build GRCh38
rs774083204 Chr.22: 50571077-50571077 on Build GRCh38
rs868688112 Chr.22: 50571181-50571181 on Build GRCh38
rs766819121 Chr.22: 50571202-50571202 on Build GRCh38
rs752352970 Chr.22: 50571091-50571091 on Build GRCh38
rs142545174 Chr.22: 50571194-50571194 on Build GRCh38
rs771940517 Chr.22: 50571171-50571171 on Build GRCh38
rs748040203 Chr.22: 50571207-50571207 on Build GRCh38
rs755318861 Chr.22: 50571204-50571204 on Build GRCh38
rs770881682 Chr.22: 50571091-50571094 on Build GRCh38
rs757823432 Chr.22: 50571123-50571123 on Build GRCh38
rs376411523 Chr.22: 50571047-50571047 on Build GRCh38
rs748234740 Chr.22: 50571160-50571160 on Build GRCh38
rs201093489 Chr.22: 50571124-50571124 on Build GRCh38
rs139313667 Chr.22: 50571131-50571132 on Build GRCh38
rs369704504 Chr.22: 50571136-50571136 on Build GRCh38
rs769210121 Chr.22: 50571049-50571049 on Build GRCh38
rs768981014 Chr.22: 50571159-50571159 on Build GRCh38
rs144863640 Chr.22: 50571195-50571195 on Build GRCh38
rs751857890 Chr.22: 50571203-50571203 on Build GRCh38
rs758805208 Chr.22: 50571201-50571201 on Build GRCh38
rs1054226818 Chr.22: 50571063-50571063 on Build GRCh38
rs899626519 Chr.22: 50571076-50571076 on Build GRCh38
rs753329152 Chr.22: 50571121-50571121 on Build GRCh38
rs1017847866 Chr.22: 50571167-50571167 on Build GRCh38
rs759279643 Chr.22: 50571170-50571170 on Build GRCh38
rs377156417 Chr.22: 50571172-50571172 on Build GRCh38
rs760213715 Chr.22: 50571118-50571118 on Build GRCh38
rs780172543 Chr.22: 50571141-50571141 on Build GRCh38
rs746961011 Chr.22: 50571058-50571058 on Build GRCh38
rs775418955 Chr.22: 50571241-50571241 on Build GRCh38

View Full Product Details