Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60451773-60452032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APPBP2
Entrez Gene ID
10513
Gene Name
amyloid beta precursor protein binding protein 2
Gene Aliases
APP-BP2, HS.84084, PAT1
UniGene
Hs.84084
Gene Chromosome Location
Chr.17: 60443149-60526240 on Build GRCh38
Transcripts
NM_001282476.1 NM_006380.3 XM_011524198.2 XM_011524199.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3362262 c.1382A>C p.Q461P Chr.17: 60452002-60452002 on Build GRCh38
6423538 c.1411G>T p.A471S Chr.17: 60451973-60451973 on Build GRCh38
1630318 c.1504+3A>G p.? Chr.17: 60451877-60451877 on Build GRCh38
6189961 c.1378G>A p.E460K Chr.17: 60452006-60452006 on Build GRCh38
6260880 c.1489C>A p.R497R Chr.17: 60451895-60451895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772131112 Chr.17: 60451844-60451844 on Build GRCh38
rs973101487 Chr.17: 60451852-60451852 on Build GRCh38
rs774163000 Chr.17: 60451931-60451931 on Build GRCh38
rs893487147 Chr.17: 60451840-60451840 on Build GRCh38
rs148076552 Chr.17: 60451818-60451818 on Build GRCh38
rs947596689 Chr.17: 60451965-60451965 on Build GRCh38
rs545412878 Chr.17: 60451828-60451828 on Build GRCh38
rs750448982 Chr.17: 60451830-60451830 on Build GRCh38
rs771471911 Chr.17: 60451995-60451995 on Build GRCh38
rs770801236 Chr.17: 60451921-60451921 on Build GRCh38
rs777500818 Chr.17: 60451875-60451875 on Build GRCh38
rs749107575 Chr.17: 60451889-60451889 on Build GRCh38
rs970454225 Chr.17: 60451805-60451805 on Build GRCh38
rs759587878 Chr.17: 60451971-60451971 on Build GRCh38
rs142010954 Chr.17: 60451827-60451827 on Build GRCh38
rs775115797 Chr.17: 60452003-60452003 on Build GRCh38
rs565416093 Chr.17: 60451835-60451835 on Build GRCh38
rs747173768 Chr.17: 60451855-60451855 on Build GRCh38
rs758403106 Chr.17: 60451831-60451831 on Build GRCh38
rs768901924 Chr.17: 60451862-60451862 on Build GRCh38

View Full Product Details