Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51367947-51368176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDND2
Entrez Gene ID
125875
Gene Name
claudin domain containing 2
Gene Aliases
-
UniGene
Hs.720536
Gene Chromosome Location
Chr.19: 51366750-51369255 on Build GRCh38
Transcripts
NM_152353.2 XM_011526425.2 XM_011526428.2 XM_017026244.1 XM_017026245.1 XM_017026246.1 XM_017026247.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361463 c.221T>C p.V74A Chr.19: 51367975-51367975 on Build GRCh38
367875 c.181G>A p.V61M Chr.19: 51368015-51368015 on Build GRCh38
5761014 c.241C>T p.L81L Chr.19: 51367955-51367955 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771527720 Chr.19: 51368038-51368038 on Build GRCh38
rs3745402 Chr.19: 51368103-51368103 on Build GRCh38
rs199854670 Chr.19: 51368016-51368016 on Build GRCh38
rs763417933 Chr.19: 51368034-51368034 on Build GRCh38
rs185890735 Chr.19: 51368078-51368078 on Build GRCh38
rs34129256 Chr.19: 51368092-51368093 on Build GRCh38
rs759951271 Chr.19: 51368042-51368042 on Build GRCh38
rs751993787 Chr.19: 51367968-51367968 on Build GRCh38
rs374582320 Chr.19: 51368061-51368061 on Build GRCh38
rs760045153 Chr.19: 51368035-51368036 on Build GRCh38
rs181014895 Chr.19: 51368001-51368001 on Build GRCh38
rs752758697 Chr.19: 51367984-51367984 on Build GRCh38
rs777808790 Chr.19: 51367994-51367994 on Build GRCh38
rs541798566 Chr.19: 51368083-51368083 on Build GRCh38
rs756295215 Chr.19: 51367990-51367990 on Build GRCh38
rs556338299 Chr.19: 51368030-51368030 on Build GRCh38
rs761029391 Chr.19: 51368064-51368064 on Build GRCh38
rs764242094 Chr.19: 51368068-51368068 on Build GRCh38
rs776166833 Chr.19: 51368032-51368032 on Build GRCh38
rs746481949 Chr.19: 51368021-51368021 on Build GRCh38
rs777005156 Chr.19: 51368036-51368053 on Build GRCh38
rs111848288 Chr.19: 51368049-51368049 on Build GRCh38
rs575076031 Chr.19: 51368052-51368052 on Build GRCh38
rs1010332986 Chr.19: 51368113-51368113 on Build GRCh38
rs964265768 Chr.19: 51368077-51368077 on Build GRCh38
rs140994018 Chr.19: 51367975-51367975 on Build GRCh38
rs938776537 Chr.19: 51368140-51368140 on Build GRCh38
rs900399948 Chr.19: 51368092-51368092 on Build GRCh38
rs886114538 Chr.19: 51368074-51368074 on Build GRCh38
rs749220517 Chr.19: 51367999-51367999 on Build GRCh38
rs115724427 Chr.19: 51368041-51368041 on Build GRCh38
rs1040713126 Chr.19: 51368091-51368091 on Build GRCh38
rs766420239 Chr.19: 51368017-51368020 on Build GRCh38
rs545931674 Chr.19: 51368123-51368123 on Build GRCh38
rs771562132 Chr.19: 51368013-51368013 on Build GRCh38
rs558697101 Chr.19: 51368011-51368011 on Build GRCh38
rs767795968 Chr.19: 51367983-51367983 on Build GRCh38
rs996040901 Chr.19: 51368098-51368098 on Build GRCh38
rs930394474 Chr.19: 51368046-51368046 on Build GRCh38
rs754042261 Chr.19: 51368071-51368071 on Build GRCh38
rs766628417 Chr.19: 51367965-51367965 on Build GRCh38
rs778565175 Chr.19: 51368009-51368009 on Build GRCh38
rs374413392 Chr.19: 51368004-51368004 on Build GRCh38
rs1020338651 Chr.19: 51368115-51368115 on Build GRCh38
rs559925705 Chr.19: 51368101-51368101 on Build GRCh38
rs760657397 Chr.19: 51368007-51368008 on Build GRCh38
rs892934299 Chr.19: 51368104-51368104 on Build GRCh38

View Full Product Details