Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1387372-1387609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNL2
Entrez Gene ID
81669
Gene Name
cyclin L2
Gene Aliases
ANIA-6B, CCNM, CCNS, HCLA-ISO, HLA-ISO, PCEE, SB138
UniGene
Hs.515704
Gene Chromosome Location
Chr.1: 1385711-1399338 on Build GRCh38
Transcripts
NM_001320153.1 NM_001320155.1 NM_030937.4 NR_135154.1 XM_011542216.2 XM_011542219.2 XM_011542221.2 XM_017002420.1 XM_017002421.1 XM_017002422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
894964 c.1390C>T p.R464W Chr.1: 1387404-1387404 on Build GRCh38
5682323 c.1422G>T p.R474R Chr.1: 1387372-1387372 on Build GRCh38
5673351 c.1313A>G p.Y438C Chr.1: 1387481-1387481 on Build GRCh38
4987614 c.1301G>C p.R434P Chr.1: 1387493-1387493 on Build GRCh38
4394382 c.1363C>T p.P455S Chr.1: 1387431-1387431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754255381 Chr.1: 1387499-1387499 on Build GRCh38
rs1013213928 Chr.1: 1387543-1387543 on Build GRCh38
rs756451022 Chr.1: 1387411-1387411 on Build GRCh38
rs907197808 Chr.1: 1387575-1387575 on Build GRCh38
rs542661325 Chr.1: 1387586-1387586 on Build GRCh38
rs751164412 Chr.1: 1387404-1387404 on Build GRCh38
rs753126298 Chr.1: 1387590-1387592 on Build GRCh38
rs577659961 Chr.1: 1387510-1387510 on Build GRCh38
rs763667158 Chr.1: 1387403-1387403 on Build GRCh38
rs761328629 Chr.1: 1387490-1387490 on Build GRCh38
rs745818611 Chr.1: 1387463-1387463 on Build GRCh38
rs769674256 Chr.1: 1387468-1387468 on Build GRCh38
rs745788659 Chr.1: 1387566-1387566 on Build GRCh38
rs779163870 Chr.1: 1387493-1387493 on Build GRCh38
rs760134886 Chr.1: 1387493-1387494 on Build GRCh38
rs761122977 Chr.1: 1387494-1387494 on Build GRCh38
rs200361653 Chr.1: 1387432-1387432 on Build GRCh38
rs750905989 Chr.1: 1387410-1387410 on Build GRCh38
rs139865828 Chr.1: 1387393-1387393 on Build GRCh38
rs772052659 Chr.1: 1387455-1387455 on Build GRCh38
rs146203556 Chr.1: 1387436-1387436 on Build GRCh38
rs753901516 Chr.1: 1387476-1387476 on Build GRCh38
rs775467043 Chr.1: 1387474-1387474 on Build GRCh38
rs768000759 Chr.1: 1387409-1387409 on Build GRCh38
rs146517668 Chr.1: 1387488-1387488 on Build GRCh38
rs778946864 Chr.1: 1387443-1387443 on Build GRCh38
rs368716428 Chr.1: 1387507-1387507 on Build GRCh38
rs894402625 Chr.1: 1387508-1387508 on Build GRCh38
rs900102282 Chr.1: 1387487-1387487 on Build GRCh38
rs766703647 Chr.1: 1387495-1387495 on Build GRCh38
rs780425685 Chr.1: 1387430-1387430 on Build GRCh38
rs781130242 Chr.1: 1387461-1387461 on Build GRCh38
rs757699796 Chr.1: 1387405-1387405 on Build GRCh38
rs779044856 Chr.1: 1387503-1387503 on Build GRCh38
rs765700547 Chr.1: 1387574-1387580 on Build GRCh38
rs906247443 Chr.1: 1387428-1387428 on Build GRCh38
rs369182174 Chr.1: 1387391-1387391 on Build GRCh38
rs577602975 Chr.1: 1387570-1387570 on Build GRCh38
rs139107239 Chr.1: 1387453-1387453 on Build GRCh38
rs753764825 Chr.1: 1387441-1387441 on Build GRCh38
rs200891540 Chr.1: 1387536-1387536 on Build GRCh38
rs752908717 Chr.1: 1387579-1387579 on Build GRCh38
rs777748622 Chr.1: 1387529-1387529 on Build GRCh38
rs775235841 Chr.1: 1387392-1387392 on Build GRCh38
rs779982309 Chr.1: 1387569-1387569 on Build GRCh38
rs151205333 Chr.1: 1387489-1387489 on Build GRCh38
rs563275316 Chr.1: 1387568-1387568 on Build GRCh38

View Full Product Details