Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 80266814-80267063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF5
Entrez Gene ID
2250
Gene Name
fibroblast growth factor 5
Gene Aliases
HBGF-5, Smag-82, TCMGLY
UniGene
Hs.37055
Gene Chromosome Location
Chr.4: 80266588-80291017 on Build GRCh38
Transcripts
NM_004464.3 NM_033143.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3696747 c.79G>T p.A27S Chr.4: 80266903-80266903 on Build GRCh38
4996467 c.32T>C p.F11S Chr.4: 80266856-80266856 on Build GRCh38
7158919 c.112G>T p.D38Y Chr.4: 80266936-80266936 on Build GRCh38
5026175 c.183C>T p.S61S Chr.4: 80267007-80267007 on Build GRCh38
5356048 c.202G>T p.G68C Chr.4: 80267026-80267026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs33950145 Chr.4: 80266984-80266984 on Build GRCh38
rs761315195 Chr.4: 80266976-80266976 on Build GRCh38
rs778817398 Chr.4: 80266844-80266844 on Build GRCh38
rs774843971 Chr.4: 80266901-80266901 on Build GRCh38
rs746524970 Chr.4: 80266866-80266866 on Build GRCh38
rs373447055 Chr.4: 80266946-80266946 on Build GRCh38
rs376127340 Chr.4: 80266958-80266958 on Build GRCh38
rs138396845 Chr.4: 80266907-80266907 on Build GRCh38
rs762245848 Chr.4: 80266968-80266968 on Build GRCh38
rs757248800 Chr.4: 80267015-80267015 on Build GRCh38
rs772649999 Chr.4: 80266860-80266860 on Build GRCh38
rs764330809 Chr.4: 80266896-80266896 on Build GRCh38
rs373927639 Chr.4: 80267007-80267007 on Build GRCh38
rs1047247 Chr.4: 80266949-80266949 on Build GRCh38
rs756297312 Chr.4: 80266923-80266923 on Build GRCh38
rs139356908 Chr.4: 80267036-80267036 on Build GRCh38
rs200286054 Chr.4: 80266969-80266969 on Build GRCh38
rs140803064 Chr.4: 80266858-80266858 on Build GRCh38
rs766535527 Chr.4: 80266977-80266977 on Build GRCh38
rs35626275 Chr.4: 80267031-80267031 on Build GRCh38
rs753985981 Chr.4: 80266924-80266924 on Build GRCh38
rs779790827 Chr.4: 80267035-80267035 on Build GRCh38
rs1033505418 Chr.4: 80266920-80266920 on Build GRCh38
rs749054735 Chr.4: 80266836-80266836 on Build GRCh38
rs587777580 Chr.4: 80266984-80266985 on Build GRCh38
rs774523989 Chr.4: 80266966-80266966 on Build GRCh38
rs745500116 Chr.4: 80266953-80266953 on Build GRCh38
rs761260486 Chr.4: 80267040-80267040 on Build GRCh38
rs774440979 Chr.4: 80267009-80267009 on Build GRCh38
rs768153756 Chr.4: 80266910-80266910 on Build GRCh38
rs188430345 Chr.4: 80267026-80267026 on Build GRCh38
rs748007294 Chr.4: 80266846-80266846 on Build GRCh38
rs376426021 Chr.4: 80267011-80267011 on Build GRCh38
rs765498654 Chr.4: 80267000-80267000 on Build GRCh38
rs966045094 Chr.4: 80266960-80266960 on Build GRCh38
rs527879794 Chr.4: 80266930-80266930 on Build GRCh38
rs868472584 Chr.4: 80267027-80267027 on Build GRCh38
rs769541814 Chr.4: 80266959-80266959 on Build GRCh38
rs1046389550 Chr.4: 80266902-80266902 on Build GRCh38
rs750465110 Chr.4: 80266914-80266914 on Build GRCh38
rs369885510 Chr.4: 80266926-80266926 on Build GRCh38
rs977296028 Chr.4: 80267034-80267034 on Build GRCh38
rs746472054 Chr.4: 80266951-80266951 on Build GRCh38
rs751463344 Chr.4: 80267013-80267013 on Build GRCh38
rs776286396 Chr.4: 80266882-80266882 on Build GRCh38
rs1053883049 Chr.4: 80267025-80267025 on Build GRCh38
rs770531227 Chr.4: 80266881-80266881 on Build GRCh38
rs773753552 Chr.4: 80267037-80267037 on Build GRCh38
rs754753055 Chr.4: 80266842-80266842 on Build GRCh38
rs773882310 Chr.4: 80266972-80266972 on Build GRCh38
rs199858015 Chr.4: 80266952-80266952 on Build GRCh38
rs985641232 Chr.4: 80267001-80267001 on Build GRCh38
rs564748410 Chr.4: 80267003-80267003 on Build GRCh38
rs150112478 Chr.4: 80266886-80266886 on Build GRCh38
rs1028201656 Chr.4: 80266908-80266908 on Build GRCh38
rs562192078 Chr.4: 80266925-80266925 on Build GRCh38

View Full Product Details