Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75404639-75404912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS3
Entrez Gene ID
6188
Gene Name
ribosomal protein S3
Gene Aliases
S3
UniGene
Hs.546286
Gene Chromosome Location
Chr.11: 75399491-75422302 on Build GRCh38
Transcripts
NM_001005.4 NM_001256802.1 NM_001260506.1 NM_001260507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
931986 c.616G>A p.D206N Chr.11: 75404749-75404749 on Build GRCh38
2017994 c.630T>C p.I210I Chr.11: 75404763-75404763 on Build GRCh38
6070370 c.711C>T p.P237P Chr.11: 75404844-75404844 on Build GRCh38
7426151 c.595G>T p.G199C Chr.11: 75404728-75404728 on Build GRCh38
3383830 c.568C>G p.L190V Chr.11: 75404701-75404701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11546222 Chr.11: 75404793-75404793 on Build GRCh38
rs780760239 Chr.11: 75404688-75404688 on Build GRCh38
rs560757181 Chr.11: 75404754-75404754 on Build GRCh38
rs776757218 Chr.11: 75404800-75404800 on Build GRCh38
rs375851508 Chr.11: 75404882-75404882 on Build GRCh38
rs936723303 Chr.11: 75404848-75404848 on Build GRCh38
rs182461343 Chr.11: 75404694-75404694 on Build GRCh38
rs73006114 Chr.11: 75404831-75404831 on Build GRCh38
rs11546229 Chr.11: 75404730-75404730 on Build GRCh38
rs773904943 Chr.11: 75404884-75404884 on Build GRCh38
rs879020583 Chr.11: 75404870-75404870 on Build GRCh38
rs1802702 Chr.11: 75404674-75404674 on Build GRCh38
rs139803355 Chr.11: 75404832-75404832 on Build GRCh38
rs765031534 Chr.11: 75404820-75404820 on Build GRCh38
rs759369058 Chr.11: 75404801-75404801 on Build GRCh38
rs146111650 Chr.11: 75404745-75404745 on Build GRCh38
rs11546196 Chr.11: 75404847-75404847 on Build GRCh38
rs11546195 Chr.11: 75404816-75404816 on Build GRCh38
rs935808490 Chr.11: 75404888-75404888 on Build GRCh38
rs775465477 Chr.11: 75404886-75404886 on Build GRCh38
rs11546223 Chr.11: 75404788-75404788 on Build GRCh38
rs774988250 Chr.11: 75404726-75404726 on Build GRCh38
rs374596699 Chr.11: 75404866-75404866 on Build GRCh38
rs945617487 Chr.11: 75404874-75404874 on Build GRCh38
rs1015096511 Chr.11: 75404682-75404682 on Build GRCh38
rs766600331 Chr.11: 75404796-75404796 on Build GRCh38
rs112257958 Chr.11: 75404666-75404666 on Build GRCh38
rs768972119 Chr.11: 75404718-75404718 on Build GRCh38
rs11546224 Chr.11: 75404849-75404849 on Build GRCh38
rs878935892 Chr.11: 75404806-75404806 on Build GRCh38
rs780895530 Chr.11: 75404839-75404839 on Build GRCh38
rs879118101 Chr.11: 75404670-75404670 on Build GRCh38
rs1027897699 Chr.11: 75404772-75404772 on Build GRCh38
rs374046719 Chr.11: 75404842-75404842 on Build GRCh38
rs748621426 Chr.11: 75404861-75404861 on Build GRCh38
rs368681048 Chr.11: 75404676-75404676 on Build GRCh38
rs747079232 Chr.11: 75404889-75404889 on Build GRCh38
rs892599702 Chr.11: 75404872-75404872 on Build GRCh38
rs11266773 Chr.11: 75404791-75404791 on Build GRCh38
rs748676206 Chr.11: 75404736-75404736 on Build GRCh38
rs779421649 Chr.11: 75404860-75404860 on Build GRCh38

View Full Product Details