Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 88961248-88961519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KYAT3
Entrez Gene ID
56267
Gene Name
kynurenine aminotransferase 3
Gene Aliases
CCBL2, KAT3, KATIII
UniGene
Hs.481898
Gene Chromosome Location
Chr.1: 88935773-88992960 on Build GRCh38
Transcripts
NM_001008661.2 NM_001008662.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4501794 c.598C>G p.L200V Chr.1: 88961449-88961449 on Build GRCh38
212037 c.458G>A p.R153K Chr.1: 88961487-88961487 on Build GRCh38
354917 c.637C>G p.L213V Chr.1: 88961410-88961410 on Build GRCh38
354918 c.535C>G p.L179V Chr.1: 88961410-88961410 on Build GRCh38
6993140 c.556C>T p.L186F Chr.1: 88961389-88961389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761326104 Chr.1: 88961278-88961278 on Build GRCh38
rs143845439 Chr.1: 88961441-88961441 on Build GRCh38
rs763896292 Chr.1: 88961458-88961458 on Build GRCh38
rs375894384 Chr.1: 88961282-88961282 on Build GRCh38
rs114294004 Chr.1: 88961357-88961357 on Build GRCh38
rs375791260 Chr.1: 88961358-88961358 on Build GRCh38
rs147147156 Chr.1: 88961403-88961403 on Build GRCh38
rs967375320 Chr.1: 88961326-88961326 on Build GRCh38
rs372200870 Chr.1: 88961393-88961393 on Build GRCh38
rs752751117 Chr.1: 88961293-88961293 on Build GRCh38
rs775915550 Chr.1: 88961374-88961374 on Build GRCh38
rs766176872 Chr.1: 88961301-88961301 on Build GRCh38
rs148773156 Chr.1: 88961412-88961412 on Build GRCh38
rs750106480 Chr.1: 88961287-88961287 on Build GRCh38
rs12063768 Chr.1: 88961273-88961273 on Build GRCh38
rs749668471 Chr.1: 88961380-88961380 on Build GRCh38
rs753616176 Chr.1: 88961311-88961311 on Build GRCh38
rs531404360 Chr.1: 88961388-88961388 on Build GRCh38
rs761270970 Chr.1: 88961392-88961392 on Build GRCh38
rs747031188 Chr.1: 88961344-88961344 on Build GRCh38
rs6670368 Chr.1: 88961292-88961292 on Build GRCh38
rs778319109 Chr.1: 88961286-88961286 on Build GRCh38
rs1002025948 Chr.1: 88961345-88961345 on Build GRCh38
rs778017091 Chr.1: 88961405-88961405 on Build GRCh38
rs767087812 Chr.1: 88961280-88961280 on Build GRCh38
rs183255042 Chr.1: 88961472-88961472 on Build GRCh38
rs779342033 Chr.1: 88961490-88961490 on Build GRCh38
rs767531553 Chr.1: 88961355-88961356 on Build GRCh38
rs769147723 Chr.1: 88961385-88961385 on Build GRCh38
rs200236169 Chr.1: 88961285-88961285 on Build GRCh38
rs759267353 Chr.1: 88961422-88961422 on Build GRCh38
rs761763373 Chr.1: 88961351-88961351 on Build GRCh38
rs778777720 Chr.1: 88961333-88961333 on Build GRCh38
rs775520140 Chr.1: 88961364-88961364 on Build GRCh38
rs756137479 Chr.1: 88961337-88961337 on Build GRCh38
rs773897866 Chr.1: 88961276-88961276 on Build GRCh38
rs967285454 Chr.1: 88961413-88961413 on Build GRCh38
rs780151984 Chr.1: 88961479-88961479 on Build GRCh38
rs754939276 Chr.1: 88961319-88961319 on Build GRCh38
rs1059370 Chr.1: 88961431-88961431 on Build GRCh38
rs765157353 Chr.1: 88961428-88961428 on Build GRCh38
rs768931361 Chr.1: 88961485-88961485 on Build GRCh38
rs752715895 Chr.1: 88961433-88961433 on Build GRCh38
rs749600096 Chr.1: 88961483-88961483 on Build GRCh38
rs571255203 Chr.1: 88961437-88961437 on Build GRCh38

View Full Product Details