Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6600193-6600446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf26
Entrez Gene ID
79034
Gene Name
chromosome 7 open reading frame 26
Gene Aliases
-
UniGene
Hs.487511
Gene Chromosome Location
Chr.7: 6589368-6608726 on Build GRCh38
Transcripts
NM_001303039.1 NM_024067.3 XM_005249845.4 XM_011515518.2 XM_017012614.1 XM_017012615.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4852479 c.958G>A p.D320N Chr.7: 6600206-6600206 on Build GRCh38
6361161 c.1061A>T p.Q354L Chr.7: 6600309-6600309 on Build GRCh38
3882092 c.1021G>A p.A341T Chr.7: 6600269-6600269 on Build GRCh38
5757069 c.1100G>A p.C367Y Chr.7: 6600348-6600348 on Build GRCh38
1091356 c.1059G>A p.A353A Chr.7: 6600307-6600307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778109473 Chr.7: 6600331-6600331 on Build GRCh38
rs551450682 Chr.7: 6600325-6600325 on Build GRCh38
rs201320447 Chr.7: 6600389-6600389 on Build GRCh38
rs776035333 Chr.7: 6600397-6600397 on Build GRCh38
rs565936469 Chr.7: 6600269-6600269 on Build GRCh38
rs766997494 Chr.7: 6600404-6600404 on Build GRCh38
rs759894410 Chr.7: 6600391-6600391 on Build GRCh38
rs761028012 Chr.7: 6600246-6600246 on Build GRCh38
rs1049284324 Chr.7: 6600327-6600327 on Build GRCh38
rs371465516 Chr.7: 6600381-6600381 on Build GRCh38
rs34300529 Chr.7: 6600322-6600322 on Build GRCh38
rs775211657 Chr.7: 6600278-6600278 on Build GRCh38
rs765550561 Chr.7: 6600395-6600395 on Build GRCh38
rs779893902 Chr.7: 6600353-6600353 on Build GRCh38
rs753261395 Chr.7: 6600261-6600261 on Build GRCh38
rs756737331 Chr.7: 6600336-6600336 on Build GRCh38
rs537027550 Chr.7: 6600351-6600351 on Build GRCh38
rs370707927 Chr.7: 6600240-6600240 on Build GRCh38
rs769768904 Chr.7: 6600219-6600219 on Build GRCh38
rs535753549 Chr.7: 6600401-6600401 on Build GRCh38
rs907424303 Chr.7: 6600422-6600422 on Build GRCh38
rs759040263 Chr.7: 6600328-6600328 on Build GRCh38
rs775555772 Chr.7: 6600363-6600363 on Build GRCh38
rs899172561 Chr.7: 6600236-6600236 on Build GRCh38
rs766481174 Chr.7: 6600247-6600247 on Build GRCh38
rs373633469 Chr.7: 6600390-6600390 on Build GRCh38
rs202007178 Chr.7: 6600333-6600333 on Build GRCh38
rs754280330 Chr.7: 6600254-6600254 on Build GRCh38
rs774179374 Chr.7: 6600380-6600380 on Build GRCh38
rs754039723 Chr.7: 6600301-6600301 on Build GRCh38
rs200796889 Chr.7: 6600226-6600226 on Build GRCh38
rs772529840 Chr.7: 6600213-6600213 on Build GRCh38
rs529607419 Chr.7: 6600265-6600265 on Build GRCh38
rs763384992 Chr.7: 6600398-6600398 on Build GRCh38
rs768689739 Chr.7: 6600368-6600368 on Build GRCh38
rs762154872 Chr.7: 6600307-6600307 on Build GRCh38
rs912443342 Chr.7: 6600267-6600267 on Build GRCh38
rs547557922 Chr.7: 6600268-6600268 on Build GRCh38
rs370175539 Chr.7: 6600326-6600326 on Build GRCh38
rs145649038 Chr.7: 6600293-6600293 on Build GRCh38
rs749934494 Chr.7: 6600408-6600408 on Build GRCh38
rs780709474 Chr.7: 6600344-6600344 on Build GRCh38
rs779381725 Chr.7: 6600260-6600260 on Build GRCh38
rs927598113 Chr.7: 6600376-6600376 on Build GRCh38
rs140585498 Chr.7: 6600292-6600292 on Build GRCh38
rs543435007 Chr.7: 6600223-6600223 on Build GRCh38
rs958961871 Chr.7: 6600379-6600379 on Build GRCh38
rs769603866 Chr.7: 6600271-6600271 on Build GRCh38
rs148385450 Chr.7: 6600259-6600259 on Build GRCh38
rs9791619 Chr.7: 6600405-6600405 on Build GRCh38
rs762938985 Chr.7: 6600291-6600291 on Build GRCh38
rs776553618 Chr.7: 6600218-6600218 on Build GRCh38
rs764467288 Chr.7: 6600399-6600399 on Build GRCh38
rs562230783 Chr.7: 6600262-6600262 on Build GRCh38
rs746686203 Chr.7: 6600214-6600214 on Build GRCh38
rs142754845 Chr.7: 6600352-6600352 on Build GRCh38
rs775424677 Chr.7: 6600220-6600220 on Build GRCh38
rs759766057 Chr.7: 6600306-6600306 on Build GRCh38

View Full Product Details