Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 7803619-7803860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PER3
Entrez Gene ID
8863
Gene Name
period circadian clock 3
Gene Aliases
FASPS3, GIG13
UniGene
Hs.162200
Gene Chromosome Location
Chr.1: 7784285-7845181 on Build GRCh38
Transcripts
NM_001289861.1 NM_001289862.1 NM_001289863.1 NM_001289864.1 NM_016831.2 XM_005263524.3 XM_011542385.2 XM_011542390.2 XM_017002723.1 XM_017002724.1 XM_017002725.1 XM_017002726.1 XM_017002727.1 XM_017002728.1 XM_017002729.1 XM_017002730.1 XM_017002731.1 XM_017002732.1 XM_017002733.1 XM_017002734.1 XM_017002735.1 XM_017002736.1 XM_017002737.1 XM_017002738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4390401 c.1091G>T p.S364I Chr.1: 7803806-7803806 on Build GRCh38
4010014 c.979T>C p.L327L Chr.1: 7803694-7803694 on Build GRCh38
7220378 c.1051A>T p.I351L Chr.1: 7803766-7803766 on Build GRCh38
1602693 c.977-4A>G p.? Chr.1: 7803688-7803688 on Build GRCh38
1176836 c.1103C>A p.S368Y Chr.1: 7803818-7803818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761441882 Chr.1: 7803703-7803703 on Build GRCh38
rs375965833 Chr.1: 7803756-7803756 on Build GRCh38
rs745884057 Chr.1: 7803758-7803758 on Build GRCh38
rs778909457 Chr.1: 7803749-7803749 on Build GRCh38
rs749951524 Chr.1: 7803720-7803720 on Build GRCh38
rs374017079 Chr.1: 7803780-7803780 on Build GRCh38
rs767025250 Chr.1: 7803710-7803710 on Build GRCh38
rs757458058 Chr.1: 7803754-7803754 on Build GRCh38
rs149164395 Chr.1: 7803676-7803678 on Build GRCh38
rs200722810 Chr.1: 7803812-7803812 on Build GRCh38
rs775921604 Chr.1: 7803776-7803776 on Build GRCh38
rs766000822 Chr.1: 7803810-7803810 on Build GRCh38
rs936587567 Chr.1: 7803767-7803767 on Build GRCh38
rs768704848 Chr.1: 7803689-7803689 on Build GRCh38
rs761506646 Chr.1: 7803797-7803797 on Build GRCh38
rs767029988 Chr.1: 7803800-7803800 on Build GRCh38
rs772950694 Chr.1: 7803808-7803808 on Build GRCh38
rs376535940 Chr.1: 7803681-7803683 on Build GRCh38
rs1025330907 Chr.1: 7803763-7803763 on Build GRCh38
rs764460128 Chr.1: 7803682-7803682 on Build GRCh38
rs143443665 Chr.1: 7803766-7803766 on Build GRCh38
rs763259395 Chr.1: 7803664-7803664 on Build GRCh38
rs755032501 Chr.1: 7803827-7803827 on Build GRCh38
rs774805653 Chr.1: 7803753-7803753 on Build GRCh38
rs751203705 Chr.1: 7803692-7803692 on Build GRCh38
rs868049631 Chr.1: 7803730-7803730 on Build GRCh38
rs755673447 Chr.1: 7803721-7803721 on Build GRCh38
rs372610495 Chr.1: 7803714-7803714 on Build GRCh38
rs771396328 Chr.1: 7803703-7803704 on Build GRCh38
rs199945897 Chr.1: 7803832-7803832 on Build GRCh38
rs770249778 Chr.1: 7803658-7803658 on Build GRCh38
rs753830988 Chr.1: 7803732-7803732 on Build GRCh38
rs749855218 Chr.1: 7803778-7803778 on Build GRCh38
rs752481233 Chr.1: 7803833-7803833 on Build GRCh38
rs777169668 Chr.1: 7803762-7803764 on Build GRCh38
rs774739639 Chr.1: 7803785-7803785 on Build GRCh38
rs754761909 Chr.1: 7803737-7803737 on Build GRCh38
rs191016322 Chr.1: 7803809-7803809 on Build GRCh38
rs368833517 Chr.1: 7803667-7803667 on Build GRCh38
rs758231609 Chr.1: 7803834-7803834 on Build GRCh38
rs780101079 Chr.1: 7803725-7803725 on Build GRCh38
rs202109100 Chr.1: 7803663-7803663 on Build GRCh38

View Full Product Details