Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41628055-41628280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XRCC6
Entrez Gene ID
2547
Gene Name
X-ray repair cross complementing 6
Gene Aliases
CTC75, CTCBF, G22P1, KU70, ML8, TLAA
UniGene
Hs.292493
Gene Chromosome Location
Chr.22: 41621163-41664048 on Build GRCh38
Transcripts
NM_001288976.1 NM_001288977.1 NM_001288978.1 NM_001469.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
726991 c.142A>G p.M48V Chr.22: 41628177-41628177 on Build GRCh38
6038553 c.132C>A p.A44A Chr.22: 41628167-41628167 on Build GRCh38
5888494 c.83-7C>T p.? Chr.22: 41628111-41628111 on Build GRCh38
6212390 c.195+2T>G p.? Chr.22: 41628232-41628232 on Build GRCh38
479009 c.193C>A p.Q65K Chr.22: 41628228-41628228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538300003 Chr.22: 41628185-41628185 on Build GRCh38
rs746333795 Chr.22: 41628168-41628168 on Build GRCh38
rs749083864 Chr.22: 41628114-41628114 on Build GRCh38
rs774503375 Chr.22: 41628124-41628124 on Build GRCh38
rs777034808 Chr.22: 41628219-41628219 on Build GRCh38
rs757328030 Chr.22: 41628095-41628095 on Build GRCh38
rs920294126 Chr.22: 41628141-41628141 on Build GRCh38
rs777644759 Chr.22: 41628097-41628097 on Build GRCh38
rs889386075 Chr.22: 41628238-41628247 on Build GRCh38
rs761297117 Chr.22: 41628189-41628189 on Build GRCh38
rs944506740 Chr.22: 41628227-41628227 on Build GRCh38
rs745930113 Chr.22: 41628175-41628175 on Build GRCh38
rs778330534 Chr.22: 41628100-41628100 on Build GRCh38
rs555851210 Chr.22: 41628105-41628105 on Build GRCh38
rs28384713 Chr.22: 41628102-41628103 on Build GRCh38
rs373827182 Chr.22: 41628115-41628115 on Build GRCh38
rs577676183 Chr.22: 41628149-41628149 on Build GRCh38
rs983186249 Chr.22: 41628172-41628172 on Build GRCh38
rs950456250 Chr.22: 41628166-41628166 on Build GRCh38
rs764318806 Chr.22: 41628203-41628203 on Build GRCh38

View Full Product Details