Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12578805-12579032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS3
Entrez Gene ID
9249
Gene Name
dehydrogenase/reductase 3
Gene Aliases
DD83.1, RDH17, Rsdr1, SDR1, SDR16C1, retSDR1
UniGene
Hs.289347
Gene Chromosome Location
Chr.1: 12567910-12618361 on Build GRCh38
Transcripts
NM_001319225.1 NM_001324370.1 NM_004753.6 XM_006711036.1 XM_017002809.1 NR_106788.2

Gene Symbol
MIR6730
Entrez Gene ID
102466722
Gene Name
microRNA 6730
Gene Aliases
hsa-mir-6730
UniGene
Gene Chromosome Location
Chr.1: 12578957-12579023 on Build GRCh38
Transcripts
NM_001319225.1 NM_001324370.1 NM_004753.6 XM_006711036.1 XM_017002809.1 NR_106788.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1667158 c.536T>G p.L179R Chr.1: 12578880-12578880 on Build GRCh38
1284486 c.495G>T p.L165L Chr.1: 12578921-12578921 on Build GRCh38
6212551 c.579G>A p.A193A Chr.1: 12578837-12578837 on Build GRCh38
6995760 c.608C>G p.T203S Chr.1: 12578808-12578808 on Build GRCh38
5025183 c.611T>C p.L204P Chr.1: 12578805-12578805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190251702 Chr.1: 12578890-12578890 on Build GRCh38
rs781205046 Chr.1: 12578938-12578938 on Build GRCh38
rs777915818 Chr.1: 12578838-12578838 on Build GRCh38
rs756230828 Chr.1: 12578968-12578968 on Build GRCh38
rs1020472927 Chr.1: 12578979-12578979 on Build GRCh38
rs112570881 Chr.1: 12578867-12578867 on Build GRCh38
rs368694426 Chr.1: 12578945-12578945 on Build GRCh38
rs2275874 Chr.1: 12578990-12578990 on Build GRCh38
rs529741930 Chr.1: 12578828-12578828 on Build GRCh38
rs371663205 Chr.1: 12578964-12578964 on Build GRCh38
rs200199732 Chr.1: 12578880-12578880 on Build GRCh38
rs147019072 Chr.1: 12578858-12578858 on Build GRCh38
rs931488191 Chr.1: 12578954-12578954 on Build GRCh38
rs763586480 Chr.1: 12578917-12578917 on Build GRCh38
rs771732502 Chr.1: 12578991-12578991 on Build GRCh38
rs764498599 Chr.1: 12578934-12578934 on Build GRCh38
rs377237878 Chr.1: 12578971-12578971 on Build GRCh38
rs145040210 Chr.1: 12578891-12578891 on Build GRCh38
rs768696062 Chr.1: 12578980-12578980 on Build GRCh38
rs2275875 Chr.1: 12578936-12578936 on Build GRCh38
rs749031572 Chr.1: 12578975-12578975 on Build GRCh38
rs765650113 Chr.1: 12578906-12578906 on Build GRCh38
rs867569622 Chr.1: 12578927-12578927 on Build GRCh38
rs761393294 Chr.1: 12578885-12578885 on Build GRCh38
rs773977653 Chr.1: 12578881-12578881 on Build GRCh38
rs111644113 Chr.1: 12578958-12578958 on Build GRCh38
rs34283357 Chr.1: 12579003-12579004 on Build GRCh38
rs772792187 Chr.1: 12579004-12579004 on Build GRCh38
rs904973071 Chr.1: 12578935-12578935 on Build GRCh38
rs140854623 Chr.1: 12578866-12578866 on Build GRCh38
rs371914317 Chr.1: 12578846-12578846 on Build GRCh38
rs895216442 Chr.1: 12578969-12578969 on Build GRCh38
rs368643564 Chr.1: 12578837-12578837 on Build GRCh38
rs753270838 Chr.1: 12578834-12578834 on Build GRCh38
rs1128256 Chr.1: 12578831-12578831 on Build GRCh38
rs902534175 Chr.1: 12579006-12579006 on Build GRCh38
rs754304027 Chr.1: 12578825-12578825 on Build GRCh38
rs749260101 Chr.1: 12578870-12578870 on Build GRCh38
rs760356268 Chr.1: 12578904-12578904 on Build GRCh38
rs778963029 Chr.1: 12578983-12578983 on Build GRCh38
rs757291841 Chr.1: 12578937-12578937 on Build GRCh38
rs752939370 Chr.1: 12578912-12578912 on Build GRCh38

View Full Product Details