Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 4786387-4786624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP5Z1
Entrez Gene ID
9907
Gene Name
adaptor related protein complex 5 zeta 1 subunit
Gene Aliases
KIAA0415, SPG48, zeta
UniGene
Hs.558440
Gene Chromosome Location
Chr.7: 4775617-4794395 on Build GRCh38
Transcripts
NM_014855.2 XM_017012864.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6110595 c.3423G>C p.L1141F Chr.7: 4786407-4786407 on Build GRCh38
4636159 c.3419G>A p.R1140K Chr.7: 4786403-4786403 on Build GRCh38
6601822 c.3405C>T p.H1135H Chr.7: 4786389-4786389 on Build GRCh38
3832925 c.3408C>T p.L1136L Chr.7: 4786392-4786392 on Build GRCh38
5630183 c.3431C>G p.P1144R Chr.7: 4786415-4786415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77688660 Chr.7: 4786547-4786547 on Build GRCh38
rs771938137 Chr.7: 4786420-4786420 on Build GRCh38
rs569396503 Chr.7: 4786414-4786414 on Build GRCh38
rs1007135011 Chr.7: 4786534-4786534 on Build GRCh38
rs566780778 Chr.7: 4786469-4786470 on Build GRCh38
rs139945151 Chr.7: 4786557-4786557 on Build GRCh38
rs752913500 Chr.7: 4786430-4786430 on Build GRCh38
rs768481273 Chr.7: 4786415-4786415 on Build GRCh38
rs971537734 Chr.7: 4786463-4786463 on Build GRCh38
rs891856292 Chr.7: 4786475-4786475 on Build GRCh38
rs184587800 Chr.7: 4786575-4786575 on Build GRCh38
rs141712634 Chr.7: 4786500-4786500 on Build GRCh38
rs971850682 Chr.7: 4786606-4786606 on Build GRCh38
rs779765784 Chr.7: 4786452-4786452 on Build GRCh38
rs930524017 Chr.7: 4786483-4786483 on Build GRCh38
rs778733323 Chr.7: 4786469-4786469 on Build GRCh38
rs756668364 Chr.7: 4786551-4786551 on Build GRCh38
rs973450216 Chr.7: 4786553-4786553 on Build GRCh38
rs1000379643 Chr.7: 4786563-4786563 on Build GRCh38
rs569253250 Chr.7: 4786467-4786468 on Build GRCh38
rs1040260765 Chr.7: 4786528-4786528 on Build GRCh38
rs535052454 Chr.7: 4786501-4786501 on Build GRCh38
rs771622825 Chr.7: 4786600-4786600 on Build GRCh38
rs757732806 Chr.7: 4786440-4786440 on Build GRCh38
rs1029795543 Chr.7: 4786523-4786523 on Build GRCh38
rs1033204347 Chr.7: 4786592-4786592 on Build GRCh38
rs764497844 Chr.7: 4786435-4786435 on Build GRCh38
rs891422071 Chr.7: 4786526-4786526 on Build GRCh38
rs751910849 Chr.7: 4786439-4786439 on Build GRCh38
rs753088431 Chr.7: 4786427-4786427 on Build GRCh38
rs573712663 Chr.7: 4786468-4786468 on Build GRCh38
rs555185439 Chr.7: 4786441-4786441 on Build GRCh38
rs755680426 Chr.7: 4786448-4786448 on Build GRCh38
rs150562452 Chr.7: 4786518-4786518 on Build GRCh38
rs559025607 Chr.7: 4786546-4786546 on Build GRCh38
rs775284953 Chr.7: 4786407-4786407 on Build GRCh38
rs771941749 Chr.7: 4786474-4786474 on Build GRCh38
rs201200772 Chr.7: 4786433-4786433 on Build GRCh38
rs927321562 Chr.7: 4786479-4786479 on Build GRCh38
rs545369038 Chr.7: 4786561-4786561 on Build GRCh38
rs79109928 Chr.7: 4786560-4786560 on Build GRCh38
rs762849835 Chr.7: 4786442-4786472 on Build GRCh38
rs774058976 Chr.7: 4786416-4786416 on Build GRCh38
rs1024513988 Chr.7: 4786605-4786605 on Build GRCh38
rs748944609 Chr.7: 4786457-4786457 on Build GRCh38
rs760674124 Chr.7: 4786478-4786478 on Build GRCh38
rs773141544 Chr.7: 4786476-4786476 on Build GRCh38
rs761776149 Chr.7: 4786418-4786418 on Build GRCh38
rs964428474 Chr.7: 4786548-4786548 on Build GRCh38
rs1010350249 Chr.7: 4786601-4786601 on Build GRCh38
rs374901857 Chr.7: 4786445-4786445 on Build GRCh38
rs907686768 Chr.7: 4786512-4786512 on Build GRCh38
rs541987188 Chr.7: 4786513-4786513 on Build GRCh38
rs113569690 Chr.7: 4786471-4786472 on Build GRCh38
rs371537061 Chr.7: 4786425-4786425 on Build GRCh38

View Full Product Details