Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 54695591-54695834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIT
Entrez Gene ID
3815
Gene Name
KIT proto-oncogene receptor tyrosine kinase
Gene Aliases
C-Kit, CD117, PBT, SCFR
UniGene
Hs.479754
Gene Chromosome Location
Chr.4: 54657928-54740715 on Build GRCh38
Transcripts
NM_000222.2 NM_001093772.1 XM_005265740.1 XM_005265741.1 XM_005265742.2 XM_017008178.1 XM_017008179.1 XM_017008180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6983718 c.291delC p.C97fs*1 Chr.4: 54695735-54695735 on Build GRCh38
1430110 c.303C>T p.H101H Chr.4: 54695747-54695747 on Build GRCh38
6437972 c.265G>A p.A89T Chr.4: 54695709-54695709 on Build GRCh38
3301421 c.200C>G p.T67S Chr.4: 54695644-54695644 on Build GRCh38
4125146 c.158A>G p.E53G Chr.4: 54695602-54695602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745640513 Chr.4: 54695627-54695627 on Build GRCh38
rs781130745 Chr.4: 54695745-54695745 on Build GRCh38
rs750916458 Chr.4: 54695687-54695687 on Build GRCh38
rs145333060 Chr.4: 54695747-54695747 on Build GRCh38
rs144933028 Chr.4: 54695644-54695644 on Build GRCh38
rs200323511 Chr.4: 54695796-54695796 on Build GRCh38
rs780922356 Chr.4: 54695764-54695764 on Build GRCh38
rs769632130 Chr.4: 54695748-54695748 on Build GRCh38
rs773860127 Chr.4: 54695803-54695803 on Build GRCh38
rs755092278 Chr.4: 54695615-54695615 on Build GRCh38
rs1020222899 Chr.4: 54695705-54695705 on Build GRCh38
rs139441923 Chr.4: 54695643-54695643 on Build GRCh38
rs376514186 Chr.4: 54695800-54695800 on Build GRCh38
rs768599276 Chr.4: 54695741-54695741 on Build GRCh38
rs202032190 Chr.4: 54695795-54695795 on Build GRCh38
rs779652404 Chr.4: 54695761-54695761 on Build GRCh38
rs749024557 Chr.4: 54695763-54695763 on Build GRCh38
rs200121443 Chr.4: 54695639-54695639 on Build GRCh38
rs201872586 Chr.4: 54695695-54695695 on Build GRCh38
rs147943899 Chr.4: 54695665-54695665 on Build GRCh38
rs747004948 Chr.4: 54695619-54695619 on Build GRCh38
rs150026676 Chr.4: 54695732-54695732 on Build GRCh38
rs146081659 Chr.4: 54695724-54695724 on Build GRCh38
rs56411694 Chr.4: 54695696-54695696 on Build GRCh38
rs779113666 Chr.4: 54695729-54695729 on Build GRCh38
rs867439564 Chr.4: 54695730-54695730 on Build GRCh38
rs866645500 Chr.4: 54695654-54695654 on Build GRCh38
rs904550523 Chr.4: 54695725-54695725 on Build GRCh38
rs959320112 Chr.4: 54695738-54695738 on Build GRCh38
rs371353189 Chr.4: 54695674-54695674 on Build GRCh38
rs557317141 Chr.4: 54695626-54695626 on Build GRCh38
rs929276403 Chr.4: 54695799-54695799 on Build GRCh38
rs374213377 Chr.4: 54695797-54695797 on Build GRCh38
rs778802467 Chr.4: 54695618-54695618 on Build GRCh38
rs373066995 Chr.4: 54695720-54695720 on Build GRCh38
rs794726674 Chr.4: 54695697-54695697 on Build GRCh38
rs768223628 Chr.4: 54695645-54695645 on Build GRCh38
rs771673266 Chr.4: 54695802-54695802 on Build GRCh38
rs762453840 Chr.4: 54695666-54695666 on Build GRCh38

View Full Product Details