Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102834656-102834905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP17A1
Entrez Gene ID
1586
Gene Name
cytochrome P450 family 17 subfamily A member 1
Gene Aliases
CPT7, CYP17, P450C17, S17AH
UniGene
Hs.438016
Gene Chromosome Location
Chr.10: 102830531-102837533 on Build GRCh38
Transcripts
NM_000102.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6128135 c.601T>C p.Y201H Chr.10: 102834850-102834850 on Build GRCh38
5580751 c.656C>T p.P219L Chr.10: 102834795-102834795 on Build GRCh38
5958823 c.574delG p.D192fs*4 Chr.10: 102834877-102834877 on Build GRCh38
4675867 c.617T>C p.I206T Chr.10: 102834834-102834834 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1004467 Chr.10: 102834750-102834750 on Build GRCh38
rs765148467 Chr.10: 102834840-102834840 on Build GRCh38
rs769089279 Chr.10: 102834877-102834877 on Build GRCh38
rs752518511 Chr.10: 102834842-102834842 on Build GRCh38
rs142435666 Chr.10: 102834823-102834823 on Build GRCh38
rs775901707 Chr.10: 102834874-102834874 on Build GRCh38
rs200063521 Chr.10: 102834805-102834805 on Build GRCh38
rs76228098 Chr.10: 102834794-102834794 on Build GRCh38
rs375364403 Chr.10: 102834875-102834875 on Build GRCh38
rs762422304 Chr.10: 102834770-102834770 on Build GRCh38
rs767247558 Chr.10: 102834803-102834803 on Build GRCh38
rs745991883 Chr.10: 102834863-102834863 on Build GRCh38
rs776597678 Chr.10: 102834782-102834782 on Build GRCh38
rs104894150 Chr.10: 102834850-102834850 on Build GRCh38
rs558207891 Chr.10: 102834731-102834731 on Build GRCh38
rs61754273 Chr.10: 102834861-102834861 on Build GRCh38
rs760032797 Chr.10: 102834783-102834783 on Build GRCh38
rs754941252 Chr.10: 102834834-102834834 on Build GRCh38
rs770270001 Chr.10: 102834785-102834785 on Build GRCh38
rs777576356 Chr.10: 102834845-102834845 on Build GRCh38
rs747064195 Chr.10: 102834763-102834763 on Build GRCh38
rs879802265 Chr.10: 102834791-102834791 on Build GRCh38
rs375350911 Chr.10: 102834797-102834797 on Build GRCh38
rs533771699 Chr.10: 102834722-102834722 on Build GRCh38
rs750052005 Chr.10: 102834813-102834813 on Build GRCh38
rs770311570 Chr.10: 102834868-102834868 on Build GRCh38
rs556134667 Chr.10: 102834828-102834828 on Build GRCh38
rs772270950 Chr.10: 102834737-102834737 on Build GRCh38
rs775909420 Chr.10: 102834795-102834795 on Build GRCh38
rs949047540 Chr.10: 102834691-102834691 on Build GRCh38
rs778700597 Chr.10: 102834734-102834734 on Build GRCh38
rs372323467 Chr.10: 102834862-102834862 on Build GRCh38
rs575801994 Chr.10: 102834844-102834844 on Build GRCh38
rs760421916 Chr.10: 102834816-102834816 on Build GRCh38

View Full Product Details