Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50641574-50641837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPKAPK3
Entrez Gene ID
7867
Gene Name
mitogen-activated protein kinase-activated protein kinase 3
Gene Aliases
3PK, MAPKAP-K3, MAPKAP3, MAPKAPK-3, MK-3
UniGene
Hs.234521
Gene Chromosome Location
Chr.3: 50611862-50649297 on Build GRCh38
Transcripts
NM_001243925.1 NM_001243926.1 NM_004635.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5621215 c.383G>A p.S128N Chr.3: 50641730-50641730 on Build GRCh38
3824234 c.369T>G p.G123G Chr.3: 50641716-50641716 on Build GRCh38
4118794 c.403G>A p.D135N Chr.3: 50641750-50641750 on Build GRCh38
446765 c.405C>A p.D135E Chr.3: 50641752-50641752 on Build GRCh38
1424210 c.424+2T>C p.? Chr.3: 50641773-50641773 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764780352 Chr.3: 50641767-50641767 on Build GRCh38
rs770936440 Chr.3: 50641723-50641723 on Build GRCh38
rs72288687 Chr.3: 50641614-50641616 on Build GRCh38
rs533150325 Chr.3: 50641604-50641604 on Build GRCh38
rs754520339 Chr.3: 50641799-50641799 on Build GRCh38
rs369078904 Chr.3: 50641660-50641660 on Build GRCh38
rs763883941 Chr.3: 50641745-50641745 on Build GRCh38
rs761480203 Chr.3: 50641750-50641750 on Build GRCh38
rs1026307843 Chr.3: 50641646-50641646 on Build GRCh38
rs775483656 Chr.3: 50641741-50641741 on Build GRCh38
rs778148724 Chr.3: 50641794-50641794 on Build GRCh38
rs760401804 Chr.3: 50641744-50641744 on Build GRCh38
rs748495326 Chr.3: 50641676-50641676 on Build GRCh38
rs371731385 Chr.3: 50641667-50641667 on Build GRCh38
rs551562759 Chr.3: 50641638-50641638 on Build GRCh38
rs753132398 Chr.3: 50641780-50641780 on Build GRCh38
rs376480526 Chr.3: 50641722-50641722 on Build GRCh38
rs749520848 Chr.3: 50641797-50641797 on Build GRCh38
rs897069890 Chr.3: 50641609-50641609 on Build GRCh38
rs749907893 Chr.3: 50641775-50641775 on Build GRCh38
rs756463941 Chr.3: 50641786-50641786 on Build GRCh38
rs779201768 Chr.3: 50641808-50641808 on Build GRCh38
rs199541398 Chr.3: 50641734-50641734 on Build GRCh38
rs777859107 Chr.3: 50641695-50641695 on Build GRCh38
rs1033731175 Chr.3: 50641781-50641781 on Build GRCh38
rs145392030 Chr.3: 50641670-50641670 on Build GRCh38
rs753466591 Chr.3: 50641749-50641749 on Build GRCh38
rs771153536 Chr.3: 50641805-50641805 on Build GRCh38
rs755532101 Chr.3: 50641776-50641776 on Build GRCh38
rs35153704 Chr.3: 50641598-50641598 on Build GRCh38
rs534053112 Chr.3: 50641728-50641728 on Build GRCh38
rs770133992 Chr.3: 50641685-50641685 on Build GRCh38
rs781671010 Chr.3: 50641778-50641778 on Build GRCh38
rs979780091 Chr.3: 50641635-50641635 on Build GRCh38
rs6791880 Chr.3: 50641813-50641813 on Build GRCh38

View Full Product Details