Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134384078-134384337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GAT1
Entrez Gene ID
27087
Gene Name
beta-1,3-glucuronyltransferase 1
Gene Aliases
CD57, GLCATP, GLCUATP, HNK1, LEU7, NK-1, NK1
UniGene
Hs.381050
Gene Chromosome Location
Chr.11: 134378504-134411986 on Build GRCh38
Transcripts
NM_018644.3 NM_054025.2 XM_005271506.3 XM_011542751.2 XM_011542753.2 XM_017017550.1 XM_017017551.1 XM_017017552.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5617664 c.259A>C p.T87P Chr.11: 134384081-134384081 on Build GRCh38
4167555 c.211T>C p.Y71H Chr.11: 134384090-134384090 on Build GRCh38
4664876 c.200G>T p.R67M Chr.11: 134384140-134384140 on Build GRCh38
4784317 c.213G>A p.T71T Chr.11: 134384127-134384127 on Build GRCh38
3445565 c.136G>A p.E46K Chr.11: 134384165-134384165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142778413 Chr.11: 134384139-134384139 on Build GRCh38
rs755439844 Chr.11: 134384127-134384127 on Build GRCh38
rs1023578235 Chr.11: 134384295-134384295 on Build GRCh38
rs535794361 Chr.11: 134384119-134384119 on Build GRCh38
rs754184239 Chr.11: 134384177-134384177 on Build GRCh38
rs771061202 Chr.11: 134384221-134384221 on Build GRCh38
rs575703343 Chr.11: 134384310-134384310 on Build GRCh38
rs557131019 Chr.11: 134384229-134384229 on Build GRCh38
rs575319632 Chr.11: 134384148-134384148 on Build GRCh38
rs888677996 Chr.11: 134384258-134384258 on Build GRCh38
rs1055690051 Chr.11: 134384274-134384274 on Build GRCh38
rs374540626 Chr.11: 134384160-134384160 on Build GRCh38
rs866692838 Chr.11: 134384165-134384165 on Build GRCh38
rs983772762 Chr.11: 134384292-134384292 on Build GRCh38
rs139621727 Chr.11: 134384151-134384151 on Build GRCh38
rs780237150 Chr.11: 134384227-134384232 on Build GRCh38
rs201436728 Chr.11: 134384112-134384112 on Build GRCh38
rs749918478 Chr.11: 134384194-134384194 on Build GRCh38
rs773485236 Chr.11: 134384098-134384098 on Build GRCh38
rs759591807 Chr.11: 134384157-134384157 on Build GRCh38
rs747001964 Chr.11: 134384236-134384239 on Build GRCh38
rs768096424 Chr.11: 134384115-134384115 on Build GRCh38
rs747932833 Chr.11: 134384220-134384220 on Build GRCh38
rs1025394740 Chr.11: 134384275-134384275 on Build GRCh38
rs891837806 Chr.11: 134384170-134384170 on Build GRCh38
rs748933614 Chr.11: 134384126-134384126 on Build GRCh38
rs746984559 Chr.11: 134384206-134384206 on Build GRCh38
rs901303092 Chr.11: 134384250-134384250 on Build GRCh38
rs761233679 Chr.11: 134384172-134384172 on Build GRCh38
rs575105698 Chr.11: 134384117-134384117 on Build GRCh38
rs766405068 Chr.11: 134384102-134384102 on Build GRCh38
rs150624857 Chr.11: 134384144-134384144 on Build GRCh38
rs780868366 Chr.11: 134384208-134384208 on Build GRCh38
rs761287009 Chr.11: 134384236-134384236 on Build GRCh38
rs778311563 Chr.11: 134384133-134384133 on Build GRCh38
rs768509148 Chr.11: 134384145-134384145 on Build GRCh38
rs745343809 Chr.11: 134384136-134384136 on Build GRCh38
rs778268096 Chr.11: 134384124-134384124 on Build GRCh38
rs894567444 Chr.11: 134384301-134384301 on Build GRCh38
rs367583989 Chr.11: 134384111-134384111 on Build GRCh38
rs1019793586 Chr.11: 134384235-134384235 on Build GRCh38
rs763229418 Chr.11: 134384100-134384100 on Build GRCh38
rs753312357 Chr.11: 134384168-134384168 on Build GRCh38
rs764601885 Chr.11: 134384175-134384175 on Build GRCh38
rs952389558 Chr.11: 134384291-134384291 on Build GRCh38
rs564214354 Chr.11: 134384252-134384252 on Build GRCh38
rs768554519 Chr.11: 134384207-134384207 on Build GRCh38
rs778563792 Chr.11: 134384192-134384192 on Build GRCh38
rs941379897 Chr.11: 134384268-134384268 on Build GRCh38
rs368383710 Chr.11: 134384199-134384199 on Build GRCh38
rs772344058 Chr.11: 134384234-134384234 on Build GRCh38
rs756892799 Chr.11: 134384179-134384179 on Build GRCh38
rs61908688 Chr.11: 134384228-134384228 on Build GRCh38
rs748418876 Chr.11: 134384248-134384248 on Build GRCh38
rs545866474 Chr.11: 134384203-134384203 on Build GRCh38
rs776452256 Chr.11: 134384147-134384147 on Build GRCh38
rs749619638 Chr.11: 134384105-134384105 on Build GRCh38

View Full Product Details