Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31386627-31386874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSG1-AS1
Entrez Gene ID
101927718
Gene Name
DSG1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.625473
Gene Chromosome Location
Chr.18: 31343368-31426988 on Build GRCh38
Transcripts
NR_110788.1 NM_001134453.2 NM_177986.4

Gene Symbol
DSG4
Entrez Gene ID
147409
Gene Name
desmoglein 4
Gene Aliases
CDGF13, CDHF13, HYPT6, LAH
UniGene
Hs.407618
Gene Chromosome Location
Chr.18: 31376777-31415791 on Build GRCh38
Transcripts
NR_110788.1 NM_001134453.2 NM_177986.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3989524 c.95T>A p.F32Y Chr.18: 31386698-31386698 on Build GRCh38
3525073 c.87G>A p.V29V Chr.18: 31386690-31386690 on Build GRCh38
7140792 c.97G>T p.D33Y Chr.18: 31386700-31386700 on Build GRCh38
708117 c.170C>A p.A57D Chr.18: 31386773-31386773 on Build GRCh38
3525075 c.195G>C p.S65S Chr.18: 31386798-31386798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201146453 Chr.18: 31386703-31386703 on Build GRCh38
rs774203896 Chr.18: 31386781-31386781 on Build GRCh38
rs1020503126 Chr.18: 31386802-31386802 on Build GRCh38
rs778441344 Chr.18: 31386819-31386819 on Build GRCh38
rs766478431 Chr.18: 31386659-31386659 on Build GRCh38
rs149672864 Chr.18: 31386775-31386775 on Build GRCh38
rs779332288 Chr.18: 31386749-31386749 on Build GRCh38
rs755784920 Chr.18: 31386730-31386730 on Build GRCh38
rs747434090 Chr.18: 31386854-31386854 on Build GRCh38
rs766413476 Chr.18: 31386780-31386782 on Build GRCh38
rs761757747 Chr.18: 31386785-31386785 on Build GRCh38
rs370177706 Chr.18: 31386760-31386760 on Build GRCh38
rs747237901 Chr.18: 31386768-31386768 on Build GRCh38
rs1041276883 Chr.18: 31386675-31386675 on Build GRCh38
rs755839912 Chr.18: 31386799-31386799 on Build GRCh38
rs753849768 Chr.18: 31386660-31386660 on Build GRCh38
rs1011420811 Chr.18: 31386798-31386798 on Build GRCh38
rs753512705 Chr.18: 31386811-31386811 on Build GRCh38
rs758874397 Chr.18: 31386843-31386843 on Build GRCh38
rs200219673 Chr.18: 31386769-31386769 on Build GRCh38
rs776765973 Chr.18: 31386817-31386818 on Build GRCh38
rs139940348 Chr.18: 31386716-31386716 on Build GRCh38
rs763994902 Chr.18: 31386697-31386697 on Build GRCh38
rs779712549 Chr.18: 31386734-31386734 on Build GRCh38
rs748868284 Chr.18: 31386740-31386740 on Build GRCh38
rs750172755 Chr.18: 31386797-31386797 on Build GRCh38
rs938260200 Chr.18: 31386796-31386796 on Build GRCh38
rs778196375 Chr.18: 31386812-31386812 on Build GRCh38
rs372495403 Chr.18: 31386668-31386668 on Build GRCh38
rs868210970 Chr.18: 31386801-31386801 on Build GRCh38
rs374462685 Chr.18: 31386795-31386795 on Build GRCh38
rs748670675 Chr.18: 31386756-31386756 on Build GRCh38
rs751508921 Chr.18: 31386699-31386699 on Build GRCh38
rs773587069 Chr.18: 31386763-31386763 on Build GRCh38
rs766203357 Chr.18: 31386805-31386805 on Build GRCh38
rs754509456 Chr.18: 31386748-31386748 on Build GRCh38
rs760837873 Chr.18: 31386658-31386658 on Build GRCh38
rs527707261 Chr.18: 31386779-31386779 on Build GRCh38
rs552571097 Chr.18: 31386842-31386842 on Build GRCh38
rs899875809 Chr.18: 31386685-31386685 on Build GRCh38
rs778125387 Chr.18: 31386847-31386847 on Build GRCh38
rs762893825 Chr.18: 31386690-31386690 on Build GRCh38
rs991120116 Chr.18: 31386820-31386820 on Build GRCh38

View Full Product Details