Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2933095-2933348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLYWCH1
Entrez Gene ID
84256
Gene Name
FLYWCH-type zinc finger 1
Gene Aliases
-
UniGene
Hs.655321
Gene Chromosome Location
Chr.16: 2911873-2951208 on Build GRCh38
Transcripts
NM_001308068.1 NM_020912.1 NM_032296.2 XM_006720962.3 XM_011522701.2 XM_011522702.2 XM_011522703.2 XM_011522704.2 XM_011522705.2 XM_011522707.2 XM_017023772.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
372808 c.794-2A>G p.? Chr.16: 2933128-2933128 on Build GRCh38
1207281 c.1000A>G p.M334V Chr.16: 2933336-2933336 on Build GRCh38
1207283 c.919G>T p.A307S Chr.16: 2933255-2933255 on Build GRCh38
4515025 c.998C>T p.P333L Chr.16: 2933331-2933331 on Build GRCh38
1377228 c.870G>T p.K290N Chr.16: 2933206-2933206 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201661996 Chr.16: 2933254-2933254 on Build GRCh38
rs777839097 Chr.16: 2933217-2933217 on Build GRCh38
rs759880840 Chr.16: 2933187-2933187 on Build GRCh38
rs745354466 Chr.16: 2933322-2933322 on Build GRCh38
rs376283105 Chr.16: 2933221-2933221 on Build GRCh38
rs377699630 Chr.16: 2933276-2933276 on Build GRCh38
rs368607158 Chr.16: 2933161-2933161 on Build GRCh38
rs1009315640 Chr.16: 2933166-2933166 on Build GRCh38
rs372673554 Chr.16: 2933117-2933117 on Build GRCh38
rs972111083 Chr.16: 2933214-2933214 on Build GRCh38
rs374207594 Chr.16: 2933271-2933271 on Build GRCh38
rs774176037 Chr.16: 2933274-2933274 on Build GRCh38
rs770745025 Chr.16: 2933187-2933189 on Build GRCh38
rs755028500 Chr.16: 2933168-2933168 on Build GRCh38
rs373732653 Chr.16: 2933194-2933194 on Build GRCh38
rs370240422 Chr.16: 2933290-2933290 on Build GRCh38
rs769201421 Chr.16: 2933233-2933233 on Build GRCh38
rs748281841 Chr.16: 2933173-2933173 on Build GRCh38
rs781275024 Chr.16: 2933169-2933169 on Build GRCh38
rs778898120 Chr.16: 2933296-2933296 on Build GRCh38
rs181440916 Chr.16: 2933257-2933257 on Build GRCh38
rs763607368 Chr.16: 2933149-2933149 on Build GRCh38
rs761336488 Chr.16: 2933189-2933189 on Build GRCh38
rs577948988 Chr.16: 2933121-2933121 on Build GRCh38
rs373503381 Chr.16: 2933139-2933139 on Build GRCh38
rs769975621 Chr.16: 2933179-2933179 on Build GRCh38
rs753402849 Chr.16: 2933150-2933150 on Build GRCh38
rs147737914 Chr.16: 2933122-2933122 on Build GRCh38
rs553954646 Chr.16: 2933123-2933123 on Build GRCh38
rs771881187 Chr.16: 2933129-2933129 on Build GRCh38
rs760381509 Chr.16: 2933285-2933285 on Build GRCh38
rs763273334 Chr.16: 2933144-2933144 on Build GRCh38
rs764835386 Chr.16: 2933154-2933154 on Build GRCh38
rs1015426876 Chr.16: 2933199-2933199 on Build GRCh38
rs766099087 Chr.16: 2933240-2933240 on Build GRCh38
rs371092849 Chr.16: 2933147-2933147 on Build GRCh38
rs200329994 Chr.16: 2933130-2933130 on Build GRCh38
rs768884431 Chr.16: 2933313-2933313 on Build GRCh38
rs756175907 Chr.16: 2933209-2933209 on Build GRCh38
rs747444782 Chr.16: 2933312-2933312 on Build GRCh38
rs750659441 Chr.16: 2933297-2933297 on Build GRCh38
rs374956718 Chr.16: 2933256-2933256 on Build GRCh38
rs752593353 Chr.16: 2933277-2933277 on Build GRCh38
rs940770229 Chr.16: 2933133-2933133 on Build GRCh38
rs1048538609 Chr.16: 2933164-2933164 on Build GRCh38
rs772401256 Chr.16: 2933230-2933230 on Build GRCh38
rs771241826 Chr.16: 2933222-2933222 on Build GRCh38
rs369902460 Chr.16: 2933246-2933246 on Build GRCh38
rs748565715 Chr.16: 2933318-2933318 on Build GRCh38
rs749483554 Chr.16: 2933182-2933182 on Build GRCh38
rs746957252 Chr.16: 2933167-2933167 on Build GRCh38
rs993601166 Chr.16: 2933302-2933302 on Build GRCh38
rs771200219 Chr.16: 2933184-2933184 on Build GRCh38
rs199520906 Chr.16: 2933243-2933243 on Build GRCh38
rs772616446 Chr.16: 2933183-2933183 on Build GRCh38
rs773216718 Chr.16: 2933152-2933152 on Build GRCh38
rs758010941 Chr.16: 2933198-2933198 on Build GRCh38
rs554747336 Chr.16: 2933263-2933263 on Build GRCh38
rs746269743 Chr.16: 2933227-2933227 on Build GRCh38
rs1005462308 Chr.16: 2933295-2933295 on Build GRCh38
rs1054704223 Chr.16: 2933286-2933286 on Build GRCh38
rs746925373 Chr.16: 2933131-2933131 on Build GRCh38
rs370306017 Chr.16: 2933188-2933188 on Build GRCh38
rs762864130 Chr.16: 2933168-2933168 on Build GRCh38
rs766714600 Chr.16: 2933145-2933145 on Build GRCh38
rs762369669 Chr.16: 2933235-2933235 on Build GRCh38
rs377595729 Chr.16: 2933264-2933264 on Build GRCh38
rs373958193 Chr.16: 2933320-2933320 on Build GRCh38
rs117697240 Chr.16: 2933298-2933298 on Build GRCh38
rs2074363 Chr.16: 2933137-2933137 on Build GRCh38
rs372022354 Chr.16: 2933241-2933241 on Build GRCh38
rs756754287 Chr.16: 2933152-2933152 on Build GRCh38
rs576695285 Chr.16: 2933160-2933160 on Build GRCh38
rs200737177 Chr.16: 2933270-2933270 on Build GRCh38
rs367874316 Chr.16: 2933208-2933208 on Build GRCh38
rs773325166 Chr.16: 2933180-2933180 on Build GRCh38
rs776011052 Chr.16: 2933231-2933231 on Build GRCh38
rs773456091 Chr.16: 2933143-2933143 on Build GRCh38
rs373284442 Chr.16: 2933207-2933207 on Build GRCh38
rs769671434 Chr.16: 2933136-2933137 on Build GRCh38
rs372969188 Chr.16: 2933247-2933247 on Build GRCh38
rs746052226 Chr.16: 2933255-2933255 on Build GRCh38
rs774719433 Chr.16: 2933185-2933185 on Build GRCh38
rs538130265 Chr.16: 2933250-2933250 on Build GRCh38
rs1030393292 Chr.16: 2933327-2933327 on Build GRCh38
rs747322764 Chr.16: 2933258-2933258 on Build GRCh38
rs764766505 Chr.16: 2933193-2933193 on Build GRCh38
rs776798795 Chr.16: 2933138-2933138 on Build GRCh38
rs759189854 Chr.16: 2933275-2933275 on Build GRCh38
rs74005527 Chr.16: 2933146-2933146 on Build GRCh38
rs201055980 Chr.16: 2933158-2933158 on Build GRCh38
rs753694802 Chr.16: 2933294-2933294 on Build GRCh38
rs565122278 Chr.16: 2933202-2933202 on Build GRCh38
rs765938514 Chr.16: 2933200-2933200 on Build GRCh38
rs780251867 Chr.16: 2933308-2933308 on Build GRCh38
rs370537064 Chr.16: 2933316-2933316 on Build GRCh38
rs368309677 Chr.16: 2933224-2933224 on Build GRCh38
rs754016731 Chr.16: 2933251-2933251 on Build GRCh38

View Full Product Details