Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46350325-46350596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35C2
Entrez Gene ID
51006
Gene Name
solute carrier family 35 member C2
Gene Aliases
BA394O2.1, C20orf5, CGI-15, OVCOV1
UniGene
Hs.593344
Gene Chromosome Location
Chr.20: 46349528-46364461 on Build GRCh38
Transcripts
NM_001281457.1 NM_001281458.1 NM_001281459.1 NM_001281460.1 NM_015945.11 NM_173073.3 NM_173179.3 XM_011528831.1 XM_011528832.1 XM_011528833.1 XM_011528834.1 XM_011528835.1 XM_011528836.1 XM_011528837.1 XM_011528838.1 XM_017027856.1 XM_017027857.1 XM_017027858.1 XM_017027859.1 XM_017027860.1 XM_017027861.1 XM_017027862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4292635 c.1131G>A p.R377R Chr.20: 46350448-46350448 on Build GRCh38
6781668 c.1055-12_1055-9delCTCT p.? Chr.20: 46350533-46350536 on Build GRCh38
6841909 c.661G>A p.E221K Chr.20: 46350432-46350432 on Build GRCh38
3748150 c.1016T>C p.L339P Chr.20: 46350476-46350476 on Build GRCh38
4983984 c.1106A>G p.E369G Chr.20: 46350473-46350473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777282326 Chr.20: 46350434-46350434 on Build GRCh38
rs559541892 Chr.20: 46350369-46350369 on Build GRCh38
rs748135961 Chr.20: 46350361-46350361 on Build GRCh38
rs960390942 Chr.20: 46350441-46350441 on Build GRCh38
rs774814396 Chr.20: 46350383-46350383 on Build GRCh38
rs761768601 Chr.20: 46350485-46350485 on Build GRCh38
rs558153843 Chr.20: 46350420-46350421 on Build GRCh38
rs762048005 Chr.20: 46350410-46350410 on Build GRCh38
rs747917659 Chr.20: 46350543-46350544 on Build GRCh38
rs760617560 Chr.20: 46350518-46350518 on Build GRCh38
rs746200872 Chr.20: 46350384-46350384 on Build GRCh38
rs764166686 Chr.20: 46350404-46350404 on Build GRCh38
rs200593243 Chr.20: 46350513-46350513 on Build GRCh38
rs754746400 Chr.20: 46350421-46350421 on Build GRCh38
rs770509342 Chr.20: 46350377-46350377 on Build GRCh38
rs56381412 Chr.20: 46350504-46350504 on Build GRCh38
rs769911862 Chr.20: 46350503-46350503 on Build GRCh38
rs569802193 Chr.20: 46350469-46350469 on Build GRCh38
rs1010776435 Chr.20: 46350370-46350370 on Build GRCh38
rs756828575 Chr.20: 46350450-46350450 on Build GRCh38
rs767361554 Chr.20: 46350426-46350426 on Build GRCh38
rs776002355 Chr.20: 46350465-46350465 on Build GRCh38
rs765410912 Chr.20: 46350411-46350411 on Build GRCh38
rs140773064 Chr.20: 46350536-46350536 on Build GRCh38
rs963296546 Chr.20: 46350497-46350497 on Build GRCh38
rs201759159 Chr.20: 46350567-46350567 on Build GRCh38
rs200138204 Chr.20: 46350562-46350562 on Build GRCh38
rs763232543 Chr.20: 46350509-46350509 on Build GRCh38
rs756011881 Chr.20: 46350433-46350433 on Build GRCh38
rs143795303 Chr.20: 46350521-46350521 on Build GRCh38
rs766141800 Chr.20: 46350346-46350346 on Build GRCh38
rs750029416 Chr.20: 46350540-46350540 on Build GRCh38
rs370131801 Chr.20: 46350532-46350532 on Build GRCh38
rs750684096 Chr.20: 46350418-46350418 on Build GRCh38
rs778631750 Chr.20: 46350533-46350533 on Build GRCh38
rs776127176 Chr.20: 46350394-46350394 on Build GRCh38
rs768592575 Chr.20: 46350452-46350453 on Build GRCh38
rs563756287 Chr.20: 46350427-46350427 on Build GRCh38
rs746046685 Chr.20: 46350431-46350434 on Build GRCh38
rs753549951 Chr.20: 46350561-46350561 on Build GRCh38
rs201597886 Chr.20: 46350488-46350488 on Build GRCh38
rs754871054 Chr.20: 46350358-46350358 on Build GRCh38
rs761009467 Chr.20: 46350470-46350470 on Build GRCh38
rs999296893 Chr.20: 46350399-46350399 on Build GRCh38
rs144878239 Chr.20: 46350480-46350480 on Build GRCh38
rs549108249 Chr.20: 46350461-46350461 on Build GRCh38
rs774334295 Chr.20: 46350499-46350500 on Build GRCh38
rs780516172 Chr.20: 46350551-46350551 on Build GRCh38
rs761331719 Chr.20: 46350396-46350396 on Build GRCh38
rs758134648 Chr.20: 46350543-46350543 on Build GRCh38
rs370773862 Chr.20: 46350481-46350481 on Build GRCh38
rs755640907 Chr.20: 46350362-46350362 on Build GRCh38
rs202041610 Chr.20: 46350462-46350462 on Build GRCh38
rs193088910 Chr.20: 46350386-46350386 on Build GRCh38
rs752504953 Chr.20: 46350516-46350516 on Build GRCh38
rs199919922 Chr.20: 46350511-46350511 on Build GRCh38
rs531062455 Chr.20: 46350449-46350449 on Build GRCh38
rs149036444 Chr.20: 46350507-46350507 on Build GRCh38
rs369773784 Chr.20: 46350359-46350359 on Build GRCh38
rs533185342 Chr.20: 46350372-46350372 on Build GRCh38
rs751379736 Chr.20: 46350350-46350350 on Build GRCh38
rs776585681 Chr.20: 46350405-46350405 on Build GRCh38

View Full Product Details