Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32744721-32744994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAXBP1
Entrez Gene ID
94104
Gene Name
PAX3 and PAX7 binding protein 1
Gene Aliases
BM020, C21orf66, FSAP105, GCFC, GCFC1
UniGene
Hs.644004
Gene Chromosome Location
Chr.21: 32733899-32771858 on Build GRCh38
Transcripts
NM_013329.3 NM_016631.3 NR_027873.1 XM_006724066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4994280 c.2089G>A p.D697N Chr.21: 32744893-32744893 on Build GRCh38
1483912 c.2126G>C p.G709A Chr.21: 32744856-32744856 on Build GRCh38
69934 c.2158T>G p.S720A Chr.21: 32744824-32744824 on Build GRCh38
5894111 c.2155C>T p.P719S Chr.21: 32744827-32744827 on Build GRCh38
3550328 c.2091C>T p.D697D Chr.21: 32744891-32744891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753960628 Chr.21: 32744756-32744757 on Build GRCh38
rs1013192087 Chr.21: 32744808-32744808 on Build GRCh38
rs78435045 Chr.21: 32744819-32744819 on Build GRCh38
rs187154233 Chr.21: 32744772-32744772 on Build GRCh38
rs144752479 Chr.21: 32744831-32744831 on Build GRCh38
rs746341501 Chr.21: 32744854-32744854 on Build GRCh38
rs766872204 Chr.21: 32744757-32744758 on Build GRCh38
rs745633590 Chr.21: 32744816-32744818 on Build GRCh38
rs754652602 Chr.21: 32744821-32744821 on Build GRCh38
rs764372702 Chr.21: 32744753-32744753 on Build GRCh38
rs762819499 Chr.21: 32744954-32744954 on Build GRCh38
rs766261242 Chr.21: 32744960-32744960 on Build GRCh38
rs757963961 Chr.21: 32744780-32744780 on Build GRCh38
rs751069083 Chr.21: 32744964-32744964 on Build GRCh38
rs773251157 Chr.21: 32744952-32744952 on Build GRCh38
rs999025868 Chr.21: 32744857-32744857 on Build GRCh38
rs755029877 Chr.21: 32744757-32744759 on Build GRCh38
rs765277920 Chr.21: 32744770-32744770 on Build GRCh38
rs780870635 Chr.21: 32744775-32744775 on Build GRCh38
rs200949840 Chr.21: 32744757-32744757 on Build GRCh38
rs765853423 Chr.21: 32744810-32744810 on Build GRCh38
rs776572383 Chr.21: 32744757-32744757 on Build GRCh38
rs758588100 Chr.21: 32744774-32744775 on Build GRCh38
rs775747068 Chr.21: 32744877-32744877 on Build GRCh38
rs149793516 Chr.21: 32744823-32744823 on Build GRCh38
rs757546224 Chr.21: 32744835-32744835 on Build GRCh38
rs535298809 Chr.21: 32744912-32744912 on Build GRCh38
rs761733425 Chr.21: 32744947-32744947 on Build GRCh38
rs761004623 Chr.21: 32744750-32744750 on Build GRCh38
rs78287607 Chr.21: 32744767-32744768 on Build GRCh38
rs747018810 Chr.21: 32744898-32744898 on Build GRCh38
rs200022896 Chr.21: 32744948-32744948 on Build GRCh38
rs1025774418 Chr.21: 32744785-32744785 on Build GRCh38
rs761459910 Chr.21: 32744763-32744763 on Build GRCh38
rs766756039 Chr.21: 32744910-32744910 on Build GRCh38
rs773572681 Chr.21: 32744749-32744749 on Build GRCh38
rs776981285 Chr.21: 32744938-32744938 on Build GRCh38
rs779518072 Chr.21: 32744846-32744846 on Build GRCh38
rs772586140 Chr.21: 32744858-32744858 on Build GRCh38

View Full Product Details