Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176119097-176119324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXD10
Entrez Gene ID
3236
Gene Name
homeobox D10
Gene Aliases
HOX4, HOX4D, HOX4E, Hox-4.4
UniGene
Hs.123070
Gene Chromosome Location
Chr.2: 176116764-176119942 on Build GRCh38
Transcripts
NM_002148.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3570195 c.983A>G p.N328S Chr.2: 176119191-176119191 on Build GRCh38
717649 c.1005C>A p.A335A Chr.2: 176119213-176119213 on Build GRCh38
1254362 c.938G>C p.W313S Chr.2: 176119146-176119146 on Build GRCh38
4657470 c.892G>T p.E298* Chr.2: 176119100-176119100 on Build GRCh38
7158436 c.972G>T p.M324I Chr.2: 176119180-176119180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754325981 Chr.2: 176119259-176119259 on Build GRCh38
rs375226686 Chr.2: 176119257-176119257 on Build GRCh38
rs570165195 Chr.2: 176119252-176119252 on Build GRCh38
rs776443207 Chr.2: 176119217-176119217 on Build GRCh38
rs931706986 Chr.2: 176119287-176119287 on Build GRCh38
rs756628680 Chr.2: 176119231-176119241 on Build GRCh38
rs757577219 Chr.2: 176119193-176119193 on Build GRCh38
rs530229667 Chr.2: 176119215-176119215 on Build GRCh38
rs769828708 Chr.2: 176119283-176119283 on Build GRCh38
rs61070787 Chr.2: 176119286-176119286 on Build GRCh38
rs748072688 Chr.2: 176119282-176119282 on Build GRCh38
rs979953790 Chr.2: 176119205-176119205 on Build GRCh38
rs771690204 Chr.2: 176119210-176119210 on Build GRCh38
rs768144045 Chr.2: 176119216-176119216 on Build GRCh38
rs779290864 Chr.2: 176119268-176119268 on Build GRCh38
rs1043109481 Chr.2: 176119285-176119285 on Build GRCh38
rs760677821 Chr.2: 176119253-176119253 on Build GRCh38
rs144160937 Chr.2: 176119206-176119206 on Build GRCh38
rs769503768 Chr.2: 176119231-176119231 on Build GRCh38
rs758644637 Chr.2: 176119136-176119136 on Build GRCh38
rs768070742 Chr.2: 176119280-176119280 on Build GRCh38
rs33913965 Chr.2: 176119195-176119195 on Build GRCh38
rs75752744 Chr.2: 176119151-176119151 on Build GRCh38
rs104893634 Chr.2: 176119164-176119164 on Build GRCh38
rs759788651 Chr.2: 176119137-176119137 on Build GRCh38
rs756705427 Chr.2: 176119160-176119160 on Build GRCh38
rs757743019 Chr.2: 176119267-176119267 on Build GRCh38
rs758203855 Chr.2: 176119192-176119192 on Build GRCh38
rs754306920 Chr.2: 176119184-176119184 on Build GRCh38
rs759890279 Chr.2: 176119241-176119241 on Build GRCh38
rs116807022 Chr.2: 176119233-176119233 on Build GRCh38
rs1027108321 Chr.2: 176119197-176119197 on Build GRCh38
rs761758159 Chr.2: 176119221-176119221 on Build GRCh38
rs536574279 Chr.2: 176119291-176119291 on Build GRCh38
rs758113477 Chr.2: 176119277-176119277 on Build GRCh38
rs767742147 Chr.2: 176119155-176119155 on Build GRCh38
rs114746583 Chr.2: 176119278-176119278 on Build GRCh38
rs752807586 Chr.2: 176119157-176119157 on Build GRCh38
rs1050159244 Chr.2: 176119293-176119293 on Build GRCh38
rs767843035 Chr.2: 176119242-176119242 on Build GRCh38
rs145367242 Chr.2: 176119139-176119139 on Build GRCh38
rs750772082 Chr.2: 176119273-176119273 on Build GRCh38
rs778140250 Chr.2: 176119182-176119182 on Build GRCh38
rs376251268 Chr.2: 176119126-176119126 on Build GRCh38
rs78466921 Chr.2: 176119150-176119150 on Build GRCh38
rs756537008 Chr.2: 176119230-176119231 on Build GRCh38

View Full Product Details