Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12956730-12957001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GADD45GIP1
Entrez Gene ID
90480
Gene Name
GADD45G interacting protein 1
Gene Aliases
CKBBP2, CKbetaBP2, CRIF1, MRP-L59, PLINP, PLINP-1, PRG6, Plinp1
UniGene
Hs.515164
Gene Chromosome Location
Chr.19: 12954156-12957254 on Build GRCh38
Transcripts
NM_052850.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4623830 c.323T>C p.L108P Chr.19: 12956890-12956890 on Build GRCh38
3756355 c.235C>G p.P79A Chr.19: 12956978-12956978 on Build GRCh38
6239710 c.282G>A p.P94P Chr.19: 12956931-12956931 on Build GRCh38
3770136 c.305C>A p.S102* Chr.19: 12956908-12956908 on Build GRCh38
6880989 c.305C>T p.S102L Chr.19: 12956908-12956908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139904917 Chr.19: 12956984-12956984 on Build GRCh38
rs936934924 Chr.19: 12956801-12956801 on Build GRCh38
rs564060928 Chr.19: 12956958-12956958 on Build GRCh38
rs200027341 Chr.19: 12956903-12956903 on Build GRCh38
rs540101166 Chr.19: 12956980-12956980 on Build GRCh38
rs763875653 Chr.19: 12956832-12956832 on Build GRCh38
rs137887501 Chr.19: 12956932-12956932 on Build GRCh38
rs980813256 Chr.19: 12956761-12956761 on Build GRCh38
rs371844428 Chr.19: 12956948-12956948 on Build GRCh38
rs988382438 Chr.19: 12956804-12956804 on Build GRCh38
rs545589991 Chr.19: 12956940-12956940 on Build GRCh38
rs775231378 Chr.19: 12956829-12956829 on Build GRCh38
rs767412838 Chr.19: 12956850-12956850 on Build GRCh38
rs767324749 Chr.19: 12956962-12956962 on Build GRCh38
rs769038600 Chr.19: 12956837-12956837 on Build GRCh38
rs762840282 Chr.19: 12956820-12956820 on Build GRCh38
rs1041582498 Chr.19: 12956842-12956842 on Build GRCh38
rs748434909 Chr.19: 12956907-12956907 on Build GRCh38
rs769457002 Chr.19: 12956825-12956825 on Build GRCh38
rs780120256 Chr.19: 12956863-12956863 on Build GRCh38
rs778967927 Chr.19: 12956976-12956976 on Build GRCh38
rs747537886 Chr.19: 12956931-12956931 on Build GRCh38
rs899601129 Chr.19: 12956851-12956851 on Build GRCh38
rs761556952 Chr.19: 12956959-12956959 on Build GRCh38
rs759295295 Chr.19: 12956763-12956763 on Build GRCh38
rs749444452 Chr.19: 12956833-12956840 on Build GRCh38
rs755150760 Chr.19: 12956873-12956873 on Build GRCh38
rs542386078 Chr.19: 12956854-12956854 on Build GRCh38
rs140839998 Chr.19: 12956978-12956978 on Build GRCh38
rs775306266 Chr.19: 12956776-12956776 on Build GRCh38
rs530103425 Chr.19: 12956830-12956830 on Build GRCh38
rs553505113 Chr.19: 12956756-12956756 on Build GRCh38
rs778382283 Chr.19: 12956927-12956927 on Build GRCh38
rs914110429 Chr.19: 12956822-12956822 on Build GRCh38
rs367606437 Chr.19: 12956831-12956831 on Build GRCh38
rs774093230 Chr.19: 12956955-12956955 on Build GRCh38
rs780152098 Chr.19: 12956825-12956826 on Build GRCh38
rs754023068 Chr.19: 12956870-12956870 on Build GRCh38
rs764998749 Chr.19: 12956765-12956765 on Build GRCh38
rs751301103 Chr.19: 12956837-12956837 on Build GRCh38
rs768175172 Chr.19: 12956952-12956952 on Build GRCh38
rs755133440 Chr.19: 12956975-12956975 on Build GRCh38
rs750304726 Chr.19: 12956963-12956963 on Build GRCh38
rs888105931 Chr.19: 12956983-12956983 on Build GRCh38
rs772601670 Chr.19: 12956910-12956910 on Build GRCh38
rs750392613 Chr.19: 12956852-12956852 on Build GRCh38
rs924961691 Chr.19: 12956754-12956754 on Build GRCh38
rs766367899 Chr.19: 12956968-12956968 on Build GRCh38
rs183050762 Chr.19: 12956840-12956840 on Build GRCh38

View Full Product Details