Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 169780411-169780650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYNN
Entrez Gene ID
55892
Gene Name
myoneurin
Gene Aliases
OSZF, SBBIZ1, ZBTB31, ZNF902
UniGene
Hs.507025
Gene Chromosome Location
Chr.3: 169772247-169789716 on Build GRCh38
Transcripts
NM_001185118.1 NM_001185119.1 NM_018657.4 NR_033702.1 NR_033703.1 XM_005247621.4 XM_005247622.3 XM_005247624.3 XM_011512987.1 XM_017006864.1 XM_017006865.1 XM_017006866.1 XM_017006867.1 XM_017006868.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6883813 c.1096G>A p.G366R Chr.3: 169780625-169780625 on Build GRCh38
4795295 c.1116G>A p.Q372Q Chr.3: 169780645-169780645 on Build GRCh38
6097017 c.1098A>G p.G366G Chr.3: 169780627-169780627 on Build GRCh38
6097016 c.1098A>G p.G366G Chr.3: 169780627-169780627 on Build GRCh38
4795294 c.1116G>A p.Q372Q Chr.3: 169780645-169780645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056339417 Chr.3: 169780518-169780518 on Build GRCh38
rs751410436 Chr.3: 169780580-169780580 on Build GRCh38
rs776532751 Chr.3: 169780540-169780540 on Build GRCh38
rs569506072 Chr.3: 169780474-169780474 on Build GRCh38
rs912251869 Chr.3: 169780569-169780569 on Build GRCh38
rs766417933 Chr.3: 169780483-169780484 on Build GRCh38
rs767228827 Chr.3: 169780620-169780620 on Build GRCh38
rs771911644 Chr.3: 169780562-169780562 on Build GRCh38
rs759319161 Chr.3: 169780612-169780612 on Build GRCh38
rs762133646 Chr.3: 169780541-169780541 on Build GRCh38
rs923975864 Chr.3: 169780463-169780463 on Build GRCh38
rs765421865 Chr.3: 169780554-169780554 on Build GRCh38
rs745605259 Chr.3: 169780561-169780561 on Build GRCh38
rs530294731 Chr.3: 169780538-169780538 on Build GRCh38
rs1000747482 Chr.3: 169780526-169780526 on Build GRCh38
rs781759879 Chr.3: 169780452-169780452 on Build GRCh38
rs956658958 Chr.3: 169780530-169780530 on Build GRCh38

View Full Product Details