Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137110529-137110766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP7
Entrez Gene ID
29952
Gene Name
dipeptidyl peptidase 7
Gene Aliases
DPP2, DPPII, QPP
UniGene
Hs.37916
Gene Chromosome Location
Chr.9: 137110540-137117980 on Build GRCh38
Transcripts
NM_013379.2 XM_005266075.3 XM_006717083.3 XM_011518599.2 XM_011518600.1 XM_017014651.1 XM_017014652.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6384069 c.1388A>G p.K463R Chr.9: 137110739-137110739 on Build GRCh38
5711238 c.1393G>A p.E465K Chr.9: 137110734-137110734 on Build GRCh38
3088642 c.1461delG p.R489fs*>4 Chr.9: 137110666-137110666 on Build GRCh38
4679282 c.1453C>A p.R485S Chr.9: 137110674-137110674 on Build GRCh38
5979828 c.1393G>C p.E465Q Chr.9: 137110734-137110734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753500482 Chr.9: 137110620-137110620 on Build GRCh38
rs1040290823 Chr.9: 137110734-137110734 on Build GRCh38
rs1128125 Chr.9: 137110651-137110651 on Build GRCh38
rs376671686 Chr.9: 137110694-137110694 on Build GRCh38
rs199703007 Chr.9: 137110716-137110716 on Build GRCh38
rs763299970 Chr.9: 137110673-137110673 on Build GRCh38
rs370553153 Chr.9: 137110613-137110613 on Build GRCh38
rs372620848 Chr.9: 137110746-137110746 on Build GRCh38
rs758118726 Chr.9: 137110725-137110725 on Build GRCh38
rs779264489 Chr.9: 137110673-137110686 on Build GRCh38
rs148567809 Chr.9: 137110720-137110720 on Build GRCh38
rs768961460 Chr.9: 137110747-137110747 on Build GRCh38
rs1026382575 Chr.9: 137110588-137110588 on Build GRCh38
rs747876375 Chr.9: 137110642-137110642 on Build GRCh38
rs201310547 Chr.9: 137110617-137110617 on Build GRCh38
rs780305582 Chr.9: 137110744-137110744 on Build GRCh38
rs763211812 Chr.9: 137110607-137110607 on Build GRCh38
rs149442204 Chr.9: 137110671-137110671 on Build GRCh38
rs1128134 Chr.9: 137110581-137110581 on Build GRCh38
rs911049720 Chr.9: 137110666-137110666 on Build GRCh38
rs887893307 Chr.9: 137110597-137110597 on Build GRCh38
rs778401773 Chr.9: 137110721-137110721 on Build GRCh38
rs770008665 Chr.9: 137110658-137110658 on Build GRCh38
rs771813924 Chr.9: 137110725-137110727 on Build GRCh38
rs369123235 Chr.9: 137110631-137110631 on Build GRCh38
rs775845198 Chr.9: 137110600-137110600 on Build GRCh38
rs371121133 Chr.9: 137110708-137110708 on Build GRCh38
rs556718103 Chr.9: 137110720-137110722 on Build GRCh38
rs1055953440 Chr.9: 137110574-137110574 on Build GRCh38
rs943064207 Chr.9: 137110727-137110727 on Build GRCh38
rs549754241 Chr.9: 137110741-137110741 on Build GRCh38
rs201321992 Chr.9: 137110745-137110745 on Build GRCh38
rs772091711 Chr.9: 137110644-137110644 on Build GRCh38
rs774827653 Chr.9: 137110678-137110678 on Build GRCh38
rs764526435 Chr.9: 137110609-137110609 on Build GRCh38
rs543797975 Chr.9: 137110723-137110723 on Build GRCh38
rs778796682 Chr.9: 137110632-137110632 on Build GRCh38
rs777657528 Chr.9: 137110726-137110726 on Build GRCh38
rs11391519 Chr.9: 137110665-137110666 on Build GRCh38
rs1128115 Chr.9: 137110728-137110728 on Build GRCh38
rs374355556 Chr.9: 137110742-137110742 on Build GRCh38
rs1128129 Chr.9: 137110629-137110629 on Build GRCh38
rs868265041 Chr.9: 137110664-137110665 on Build GRCh38
rs199555646 Chr.9: 137110610-137110610 on Build GRCh38
rs926050303 Chr.9: 137110724-137110724 on Build GRCh38
rs777851739 Chr.9: 137110645-137110645 on Build GRCh38
rs114650680 Chr.9: 137110585-137110585 on Build GRCh38
rs565823684 Chr.9: 137110653-137110653 on Build GRCh38
rs976700100 Chr.9: 137110680-137110680 on Build GRCh38
rs983961024 Chr.9: 137110637-137110637 on Build GRCh38
rs144785203 Chr.9: 137110695-137110695 on Build GRCh38
rs569748528 Chr.9: 137110571-137110571 on Build GRCh38
rs768712654 Chr.9: 137110674-137110674 on Build GRCh38
rs779736474 Chr.9: 137110619-137110619 on Build GRCh38
rs558069436 Chr.9: 137110711-137110711 on Build GRCh38
rs1014500817 Chr.9: 137110586-137110586 on Build GRCh38
rs753302138 Chr.9: 137110717-137110717 on Build GRCh38
rs746948997 Chr.9: 137110737-137110737 on Build GRCh38
rs767888757 Chr.9: 137110615-137110615 on Build GRCh38

View Full Product Details